accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 99607-70-2

IUPAC Name: heptan-2-yl 2-[(5-chloroquinolin-8-yl)oxy]acetate
Synonyms: cloquintocet-mexyl
Molecular Weight (g/mol): 335.83
Molecular Formula: C18H22ClNO3
InChi Key: COYBRKAVBMYYSF-UHFFFAOYNA-N
SMILES: CCCCCC(C)OC(=O)COC1=C2N=CC=CC2=C(Cl)C=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results