accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CH3CCH3OONH(R)H2N

CAS RN 99724-19-3

IUPAC Name: (3R)-3-{[(tert-butoxy)carbonyl]amino}pyrrolidin-1-ium
Synonyms: (3R)-3-[(tert-butoxycarbonyl)amino]pyrrolidin-1-ium
Molecular Weight (g/mol): 187.26
Molecular Formula: C9H19N2O2
InChi Key: DQQJBEAXSOOCPG-SSDOTTSWSA-O
SMILES: CC(C)(C)OC(=O)N[C@@H]1CC[NH2+]C1
Molecular Weight (g/mol) 
  • (1)
  • (4)
Percent Purity 
  • (2)
  • (1)
  • (2)
Quantity 
  • (2)
  • (1)
  • (2)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (4)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)