accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 998-93-6

IUPAC Name: 4-bromoheptane
Synonyms: 4-bromoheptane
Molecular Weight (g/mol): 179.1
Molecular Formula: C7H15Br
InChi Key: BNUTXEKPXPZAIT-UHFFFAOYSA-N
SMILES: CCCC(Br)CCC
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)