accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 999-78-0

IUPAC Name: 4,4-dimethylpent-2-yne
Synonyms: 4,4-dimethylpent-2-yne
Molecular Weight (g/mol): 96.17
Molecular Formula: C7H12
InChi Key: FOALCTWKQSWRST-UHFFFAOYSA-N
SMILES: CC#CC(C)(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results