accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 999-82-6

IUPAC Name: (2-bromoethyl)phosphonic acid
Synonyms: 2-bromoethylphosphonic acid
Molecular Weight (g/mol): 188.95
Molecular Formula: C2H6BrO3P
InChi Key: BFKXVERGWXHHIH-UHFFFAOYSA-N
SMILES: OP(O)(=O)CCBr
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results