accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H3COOOOH3C

CAS RN 99909-62-3

IUPAC Name: methyl 2-oxo-5-(propan-2-yl)-2H-cyclohepta[b]furan-3-carboxylate
Synonyms: methyl 5-isopropyl-2-oxocyclohepta[b]furan-3-carboxylate
Molecular Weight (g/mol): 246.26
Molecular Formula: C14H14O4
InChi Key: HOVPKDFNWCKKGB-UHFFFAOYSA-N
SMILES: COC(=O)C1=C2C=C(C=CC=C2OC1=O)C(C)C
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results