UserName
Viewing profile as user:
2026, Fall Winter Technology
Inspired by Plants, Scientists Develop a Battery-Free Device to Produce Clean Fuel
September 2026 | 3 min read | By: Joisey Thekkumpuram Jojan
Inspired by Plants, Scientists Develop a Battery-Free Device to Produce Clean Fuel
Have you ever wondered how a tiny green plant can make its own food using nothing but sunlight, water, and air? It might sound like magic, but it's a natural process called photosynthesis. Plants use the sun's energy to turn water and carbon dioxide (the gas we exhale) into the food they need to grow.
Now imagine if we could borrow that same process—not to grow leaves, but to make clean fuel.
Learning from Nature’s Energy Factory
That's exactly what researchers at Osaka Metropolitan University have been working on. They created a battery-free artificial photosynthesis system that uses sunlight, water, and carbon dioxide to produce a liquid fuel. The new device produces formic acid, a chemical that can serve as both a fuel and an energy storage medium, offering a promising renewable energy carrier. This exciting breakthrough could make renewable energy simpler, more affordable, and more sustainable, bringing us one step closer to a cleaner future.
How a Self-Regulating System Solves the Sunlight Problem
One of the biggest challenges in artificial photosynthesis has been dealing with how variable sunlight is. As clouds pass or daylight fades, solar panels produce less electricity. Most existing systems rely on batteries or sophisticated electronic controllers to maintain stable fuel production, adding cost and complexity to the technology.
To overcome the challenge of fluctuating sunlight, a team led by Yasuo Matsubara and Yutaka Amao at the Research Center for Artificial Photosynthesis, Osaka Metropolitan University, collaborated with Iida Group Holdings Co., Ltd. to redesign the electrolyzer—the device that converts electricity into chemical energy. Instead of relying on batteries or external control devices, the researchers engineered the electrolyzer to regulate itself as sunlight conditions change.
"As sunlight increases, the electrolyzer naturally heats up. The system is designed so that this warming causes the electrical resistance to drop, allowing electricity to flow more freely," Amao explained. "This makes the system automatically adjust its electrical behavior."
Such self-regulation, “Helps keep fuel production more stable throughout the day and automates the system, while reducing dependence on batteries and costly external components," added Amao. By eliminating the need for batteries and complex electronic controls, the innovation simplifies the overall design, lowers costs, and makes artificial photosynthesis systems more practical for future large-scale production of clean fuels.
Turning Carbon Dioxide into Useful Fuel
The fuel produced by the system is formic acid, an energy-dense liquid that can be stored, transported, and later converted into electricity or used in industrial processes. Because it is made using carbon dioxide captured from the atmosphere or other sources, the process also helps recycle greenhouse gases into something useful.
Powering Tomorrow with Artificial Photosynthesis
Nature has spent billions of years perfecting photosynthesis. By learning from that process, scientists are designing technologies that could reshape how the world produces and stores energy. This innovative system brings us closer to a future where sunlight powers our homes and produces clean fuels that help reduce carbon emissions and dependence on fossil resources.
Joisey Thekkumpuram Jojan is a Thermo Fisher Scientific staff writer.
Discussion Questions
  1. How does the self-regulating electrolyzer help maintain stable fuel production when sunlight levels change?
  2. What are the advantages of producing formic acid from carbon dioxide compared with using fossil fuels?