Fisher Science Education Lab Tools
The only laboratory measurement instruments that are always cost effective.
Quickly and easily make labels for chemicals, bottles, shelves and equipment.
- Self-adhesive colored tape sticks to glass, plastic, metal, rubber and wood
- Nine millimeter (0.375 in.) wide tape
- Raised white letters readable from 8 ft.
- Tape cartridges available in black, red, blue or green
Fisher Scientific™ Counter Pens™ are the ideal cell growth colony counter.
- Easy-to-read LCD counts to 99,999
- Combination marker and counter
- Durable ABS plastic construction
- Includes battery and pen cap
Fisherbrand™ Adjustable-Spray Mini-Wash Bottles offer zero contamination and no-effort pumping.
- 240mL
- Chemical resistant
- Unbreakable bottle
- Polypropylene sprayer
- Squat shape that takes up little bench space
The unique Fisher Scientific™ Traceable™ SpatulaBalance™ scoops material and displays weight.
- Weighing range from 0 to 300.0g and 0 to 10.580 oz.
- Readability of 0.1g (0.005 oz.)
- Accuracy within ±0.2g or ±0.25%
- Graduated markings from 5 to 30mL in 5mL increments
- Detachable scoop for dishwasher-safe cleaning
Fisher Science Education Traceable Digital Calipers
The Fisher Science Education™ Traceable™ Digital Calipers are ideal for measuring inside and outside diameter.
- Designed for the lab
- Measures in inches and millimeters
- Measures rubber stoppers, vials, caps, tubing and corks
- Instant and accurate readings
- Nonconductive carbon fiber
- Includes carrying case and battery
Catalog No. | Material | Measuring Range | Resolution | Accuracy | Depth Bar | Thumb Roller/Screw Lock | Non-metallic/Non-conductive |
---|---|---|---|---|---|---|---|
Stainless steel | 150mm (6in.) | 0.01mm (0.0005in.) | ±0.03mm (±0.001in.) | Yes | Yes | No | |
Stainless steel | 200mm (8in.) | 0.01mm (0.0005in.) | ±0.03mm (±0.001in.) | Yes | Yes | No | |
Carbon fiber | 100mm (4in.) | 0.1mm (0.01in.) | ±0.2mm (±0.01in.) | No | No | Yes | |
Carbon fiber | 150mm (6in.) | 0.1mm (0.01in.) | ±0.2mm (±0.01in.) | No | No | Yes |