EXCLUSIVE: Biocartis Idylla
Streamlined Testing
The fully automated, PCR-based Biocartis™ Idylla™ System offers fast and easy access to molecular biomarker information, anywhere and anytime.
Faster Results
Only two minutes of hands-on time. Less than 150 minutes of test time. No need to send the sample to another lab or make your patients wait days for results — just insert your sample and walk away.
Less Sample
Detect up to 30 molecular targets from just one slice of biopsy tissue or a sample size of just 1mL for plasma-based assays.
Simple Workflow
The Idylla system takes you from start to finish with only a handful of steps.
Hassle-Free Operation
Any lab technician can use the Idylla system, and the compact design and scalable setup make it the right choice for any lab.
Modular Design
A single console can be used with up to eight separately operating instruments, which makes the Idylla System scalable to meet your lab's changing workload.
All-in-One Cartridges
Each sample is tested using a simple, disposable, liquid-tight cartridge that greatly reduces the risk of contamination and includes everything needed to prepare and analyze your specimen.