UserName
Viewing profile as user:
Blood Collection Needles
Blood Collection Needles
Blood collection needles are designed to be used with blood tube holders for collecting specimens in evacuated tubes or blood culture media bottles. When drawing blood from a patient, the phlebotomist attaches the needle to a holder — one end of the needle is inserted into the patient’s vein and the other end is injected into the tube stopper to access the vacuum.
Blood collection needles are intended to be used only once, and they are designed for collecting single or multiple tubes of blood. Most have a rubber sheath over the back of the needle to prevent blood from flowing into the holder during tube changes.
Find blood draw needles in your choice of style, gauge, and length.
Featured Products