Boekel Scientific Platelet Agitator and Platelet Incubators
The Boekel™ Scientific platelet incubator and agitator are specifically designed to support the short-term storage requirements of hospital blood banks, blood centers and clinical research laboratories for blood platelets utilized for human therapeutic use based on AABB recommendations.
The latest advancements in platelet storage solutions for hospital blood banks, blood centers and clinical research laboratories.
Features & Benefits:
- Inventory management system: reduces the likelihood of platelets expiring; saves time
- Uniform agitation: ensures no platelet clumping and maximum visibility of platelets
- Extra security: the alarm display has a battery backup, protecting platelets
- Thermoelectric heating and cooling system: reduced maintenance cost
- On-board thermal printer: for quick, easy temperature record keeping
- Two-year warranty included: reduced maintenance cost
- Optimizes use of bench space: less space required