Thermo Scientific Liquid Handling Solutions
Featured Products
With a history of innovation, the Thermo Scientific™ portfolio of liquid handling products continues to offer breakthrough technology.
My Pipette Creator App
Use the My Pipette Creator app to manage and share pipette programs from the comfort of your computer. Save time, reduce errors, and increase accuracy and reproducibility when you send programs to your Thermo Scientific™ E1-ClipTip™ Pipettes. Create programs or download pre-programmed protocols for specific qPCR and sample prep assay kits, then transfer the programs directly to your pipette via Bluetooth™ or micro-USB.
Learn More
F1-ClipTip Pipetting System
The new Thermo Scientific™ F1-ClipTip™ Manual Pipetting System features innovative, interlocking technology that allows you to feel the tips lock firmly in place.
Thermo Scientific F1-ClipTip Manual Pipetting System
Help ensure reproducible results and a secure seal on every channel with the Thermo Scientific F1-ClipTip Manual Pipetting System. Minimal force is needed for tip attachment and ejection. Compatible with Thermo Scientific™ ClipTip™ Pipette Tips.
Thermo Scientific Finnpipette F1 Pipetting System
The Thermo Scientific™ Finnpipette™ F1 Pipetting System is remarkably lightweight and designed with increased air boost for efficient delivery of microvolumes. For best results, use with Thermo Scientific™ Finntip™ Pipette Tips.
Thermo Scientific Finnpipette F2 Pipetting System
For accurate microvolume dispensing, choose the Thermo Scientific Finnpipette F2 Pipetting System. These pipettes are designed with a large volume display and are fully autoclavable. For best results, use with Thermo Scientific Finntip or Thermo Scientific Finntip™ Flex™ Pipette Tips.
Electronic Pipetting Systems
Thermo Scientific electronic pipettes and tip systems help blend the control and lightweight design of manual pipettes with the precision and accuracy of electronic pipettes to boost your productivity.
Thermo Scientific™ E1-ClipTip™ Electronic Pipettes enable efficient, centralized programming and sharing of protocols between pipettes and colleagues, including access to pre-programmed protocols for many of your favorite reagent kits — all from the comfort of your computer. Learn how to connect your pipette to the My Pipette Creator App by watching the video “Thermo Scientific My Pipette Creator” under More Resources.
Equalizer Challenge #1: Tubes to 96-Well Plate
Find out how to transfer lab samples effiently with Thermo Scientific™ E1-ClipTip™ Equalizer Pipettes.
Thermo Scientific E1-ClipTip Electronic Pipettes
The Thermo Scientific E1-ClipTip Pipetting System features a unique interlocking design for secure seals, index finger pipetting, and electronic tip ejection. Equalizer models allow quick and efficient sample transfers between virtually any tube, rack, microplate, or horizontal gel box.
Thermo Scientific Finnpipette Novus Electronic Pipettes
The Thermo Scientific Finnpipette Novus Electronic Pipetting System offers single-finger pipetting operation in single and multichannel models. For best results, use with Finntip or Finntip Flex Tips.
Manual and Robotic Pipette Tips
Improving Pipette Performance and Results
Choose from a wide range of features to find the best pipette tips for your needs, including low retention, extended length, wide bore, and gel loading. The Thermo Scientific brand offers tips to fit nearly every pipette and automated liquid handler.
ART Pipette Tips Video
Count on the performance and reliability of Thermo Scientific™ ART™ barrier, non-filtered, and specialty tips for any application and pipette. Enjoy the complete universal tip solution with zero contamination and versatile storage along with space-saving reload towers.
Thermo Scientific ART Pipette Tips
The Thermo Scientific ART Pipette Tip line includes barrier and non-filtered tips from 10 μL to 10 mL, as well as gel-loading and wide-bore specialty products. ART tips have a self-sealing barrier that guarantees 100% protection against aerosol and liquid contamination. Every ART tip is certified RNase, DNase, ATP, and pyrogen free.
Thermo Scientific Finntip and Finntip Flex Pipette Tips
Help ensure accurate, precise pipetting with Thermo Scientific Finntip Pipette Tips that are manufactured using premium raw materials and the latest molding techniques. Reduce the amount of force needed to attach and eject tips with Thermo Scientific Finntip Flex Pipette Tips and significantly lower the risk of repetitive stress injury (RSI) while ensuring a better seal with the pipette for higher accuracy and precision.
Thermo Scientific Robotic Tips
Designed to fit over 50 workstations, these robotic tips undergo rigorous quality control testing and have a low coefficient of variation (from 1 to 3%).
Consistent, Reproducible Pipetting
Experience a breakthrough in technology that makes daily pipetting less of a chore with the Thermo Scientific™ ClipTip™ Pipetting System — it starts with interlocking pipette tips that seal in security.
Manual or electronic Thermo Scientific ClipTip Pipettes are available with fixed or adjustable volumes and in single or multichannel configurations.
Simply fill out the form to request a demonstration.
By submitting your data, you agree to our Privacy Policy and will receive emails based on your preferences.
© Thermo Fisher Scientific, Inc. All Rights Reserved.
Thermo Scientific Good Laboratory Pipetting Practices Brochure
Find information about tools and techniques that can help improve the accuracy and precision of your pipetting.
Thermo Scientific ART Pipette Tips Brochure
The Thermo Scientific ART portfolio offers a complete selection of barrier, non-filtered, and specialty tips. Learn more about Thermo Scientific ART Pipette Tips.
Thermo Scientific ClipTip Pipetting Systems Brochure
Learn about worry-free pipetting with the Thermo Scientific ClipTip Pipetting System. Read about the features of F1-ClipTip Pipettes, E1-ClipTip Pipettes, and more.
Thermo Scientific Finnpipette Manual Pipetting Systems Brochure
Learn about the features and specifications of Thermo Scientific Manual Pipetting Systems, including Finnpipette F1, Finnpipette F2, Finntip and Finntip Flex Pipette Tips, and more.
More Resources
Thermo Scientific Finnpipette Novus Electronic Pipette Brochure
Thermo Scientific Finnpipette Novus Single and Multichannel Electronic Pipettes deliver exceptional performance. Read about the intuitive programming, index-finger operation and more.
Thermo Scientific Automation Tips Brochure
Use this quick reference guide to view Thermo Scientific automation tip varieties compatible with a wide range of liquid handling instruments.
- Thermo Scientific Automation Tips Brochure (742.3 KB)
Thermo Scientific Automation Tips and Accessories Brochure
Browse the Thermo Scientific Automation Tips and Accessories brochure to read about available tips, workstations, and more.
Thermo Scientific My Pipette Creator
Boost your productivity and research results by pairing your Thermo Scientific E1-ClipTip Pipettes with the Thermo Scientific My Pipette Creator App.