One Pattern, All Life: The Groundbreaking Discovery of a Universal Temperature Curve
What if every living organism followed the same hidden rule—from tiny bacteria to towering trees? Scientists have now uncovered exactly that: a universal temperature curve that shapes how all life on Earth grows, moves, and survives. This discovery reveals that temperature influences life in a predictable, shared pattern across species. It connects biology, physics, and evolution under one unifying principle. More importantly, it could reshape our understanding of life’s limits in a rapidly warming world.
The study reveals that life on Earth, despite its diversity, follows a shared pattern when it comes to temperature.
The Discovery of a Universal Rule
Scientists from several universities collaborated to investigate whether a universal pattern governs how all living organisms respond to temperature, testing this idea using large-scale data across diverse species and biological processes.
The team compiled an extensive dataset, bringing together over 2,500 studies that examined how living organisms respond to temperature. These studies measured key biological functions, including growth, movement, metabolism, feeding behavior, and population dynamics, across a wide range of temperatures.
Altogether, the data included around 30,000 measurements spanning seven major branches of life and 39 phyla, from microbes and plants to invertebrates and vertebrates. When the researchers standardized each dataset based on a species’ optimal temperature and its upper thermal limit, the results aligned into a single pattern. They named this shared pattern the Universal Thermal Performance Curve (UTPC).
One Curve That Connects All Life
The study reveals that life on Earth, despite its diversity, follows a shared pattern when it comes to temperature. Jean-François Arnoldi, scientist and researcher in theoretical ecology at the Centre National de la Recherche Scientifique (CNRS) in France, collaborated with Andrew L. Jackson and Nicholas L. Payne from Trinity College Dublin, along with Ignacio Peralta-Maraver from the University of Granada on the investigation. According to Jackson, who is a professor in Zoology at Trinity College Dublin, this consistency can be seen across countless species and biological groups.
In the article “Scientists discover a universal temperature curve that governs all life” in ScienceDaily, Jackson said, “Across thousands of species and almost all groups of life, including bacteria, plants, reptiles, fish, and insects, the shape of the curve that describes how performance changes with temperature is very similar,” he said, as reported in the ScienceDaily article “Scientists discover a universal temperature curve that governs all life.” “However, different species have very different optimal temperatures, ranging from 5°C to 100°C [41°F to 212°F], and their performance can vary a lot depending on the measure of performance being observed and the species in question."
Previously, scientists created many different models to explain these variations. However, the new findings suggest that these models are essentially different versions of the same underlying curve; they simply have been stretched or shifted across temperature ranges. The research also reveals that an organism’s optimal temperature and the point at which heat becomes fatal are closely interconnected.
Jackson further explained that once temperatures rise above the optimum, the safe range for survival shrinks. “Whatever the species, it simply must have a smaller temperature range at which life is viable once temperatures shift above the optimum.”
The Next Frontier in Temperature Science
The scientists are now turning their attention to what comes next: testing how far this universal pattern truly extends. Payne, a marine ecologist and associate professor from Trinity College Dublin, highlighted that this shared response to temperature appears across organisms that have evolved independently over billions of years.
Despite the vast diversity of life and billions of years of evolution, the study shows that nearly all organisms are still governed by the same underlying rule linking temperature to performance.
Going forward, the team plans to use the UTPC as a reference point to identify rare cases that might not follow this pattern. “The next step is to use this model as something of a benchmark to see if there are any species or systems we can find that may, subtly, break away from this pattern,” said Payne. “If we find any, we will be excited to ask why and how they do it—especially given forecasts of how our climate is likely to keep warming in the next decades.”
Ultimately, each new experiment that examines how organisms respond to temperature becomes another opportunity to test this underlying rule—one that influences life everywhere, from the soil beneath our feet to the vast oceans.
Joisey Thekkumpuram Jojan is a Thermo Fisher Scientific staff writer.