Heidolph Chemical Tools
Heidolph Chem Tools Catalog
Learn more about the Heidolph line of chemistry lab products, all of which are designed to optimize your flexibility, efficiency, and performance.
Featured Products
Heidolph™ Hei-Vap™ Rotary Evaporators
- Heating bath reaches a temperature of 210°C
- Rotation speed of 20-280rpm
Heidolph™ Hei-TORQUE™ Series Laboratory Overhead Stirrers
- Constant speed even under changing loads
- Safe start and stop via slide touch panel to avoid unintended stirring
Heidolph™ MR Series Magnetic Stirrer Hotplates
- For smooth to intense mixing and heating of low-viscosity fluids
- Hotplate reaches a maximum temperature of 300°C
Heidolph™ Radleys Findenser™ Super Air Condensers
- Can be used for most common chemistry applications
- For solvent volumes from 5mL to 1L