Filtered Search Results
Agilent BioTek Synergy HTX Multi-Mode Microplate Reader
Greener Choice
Offers one or more environmental benefits itemized in the U.S. FTC Green Guides.
Learn More
Offers one or more environmental benefits itemized in the U.S. FTC Green Guides.
Learn More
The BioTek Synergy HTX is a compact, economical multi-mode reader for 6- to 384-well microplates and Take3 microvolume plates. For Research Use Only. Not for use in diagnostic procedures.
| Width (English) | 16 in. |
|---|---|
| Shaking | Linear, Orbital |
| Software Type | Gen5 Microplate Reader and Imager Software,Gen5 Secure for 21 CFR Part 11 compliance (option) |
| Absorbance Range | 200 to 999 nm, 1 nm increments |
| Light Source | Xenon Flash,Tungsten Halogen (Alpha Detection/Fluorescence Intensity) |
| Width (Metric) | 40.6 cm |
| USB Connections | 2 |
| Detector Type | Photodiode,PMT |
| Reading Options | End Point,Kinetic,Spectral Scanning,Well Area Scanning |
| Accuracy | Absorbance: ±2 nm; <1% at 2.0 OD, <3% at 3.0 OD; Dispense: ±1 μL or 2% |
| No. of Dispensers | 2 Syringe Pumps |
| Voltage | 100/240 V |
| Certifications/Compliance | CE and TUV marked. RoHS compliant. |
| Weight (Metric) | 18 kg |
| Height (English) | 10 in. |
| Height (Metric) | 25.4 cm |
| Depth (English) | 15 in. |
| Incubator Temperature | 4-Zone Incubation to 50°C with Condensation Control (±0.2°C at 37°C) |
| Dynamic Range | Absorbance: 0 to 4 OD; Fluorescence Intensity/Luminesence: >6 decades |
| Product Type | Multi-Mode Microplate Reader |
| Depth (Metric) | 38 cm |
| Measurement Speed | 31 sec. (96 wells), 80 sec. (384 wells) |
| Frequency | 50/60 Hz |
| Weight (English) | 40 lb. |
| Compatibility | BioStack and Third Party Automation Compatible |
| Plate Type | 6- to 384-well Plates |
Agilent BioTek Synergy LX Multi-Mode Reader
Greener Choice
Offers one or more environmental benefits itemized in the U.S. FTC Green Guides.
Learn More
Offers one or more environmental benefits itemized in the U.S. FTC Green Guides.
Learn More
The BioTek Synergy LX multi-mode reader automates many common microplate assays. NOTE: Fluorescence filter cubes are required for fluorescence measurements (sold separately). For Research Use Only. Not for use in diagnostic procedures.
| Width (English) | 15 in. |
|---|---|
| Shaking | Linear, Orbital, Double Orbital |
| Absorbance Range | 200 to 900 nm |
| Light Source | Xenon Flash |
| For Use With (Application) | Nucleic acid and protein quantification, ELISA, BCA, Bradford and cell viability |
| Width (Metric) | 38.1 cm |
| USB Connections | 2 |
| Detector Type | Photodiode,PMT |
| Reading Options | End Point,Kinetic,Area Scanning,Absorbance Spectral Scanning |
| Filters | Monochromator Bandpass |
| Accuracy | Wavelength: ±2 nm; Optical Density: <1% at 2.0 OD, <3% at 2.5 OD |
| Voltage | 100/240 V |
| Bandwidth | ≤5 nm |
| Certifications/Compliance | CE and TUV marked. RoHS compliant. |
| Weight (Metric) | 12.3 kg |
| Warranty | 1 year |
| Depth (English) | 15 in. |
| Software Language Versions | English |
| Sensitivity | Fluorescence Intensity: Fluorescein 2 pM; Luminesence: 10 amol ATP |
| Product Type | Multi-mode Microplate Reader |
| Linearity | <1% from 0 to 2.5 OD |
| Depth (Metric) | 38.1 cm |
| Measurement Speed | 24 sec. (96 wells), 76 sec. (384 wells) |
| Frequency | 50/60 Hz |
| Weight (English) | 27 lb. |
| Power Consumption | 24 VDC power supply compatible with 100/240 VAC at 50/60 Hz, 60 W (max.) |
| Resolution | 0.001 OD |
Agilent BioTek 800 TS Absorbance Reader
Greener Choice
Offers one or more environmental benefits itemized in the U.S. FTC Green Guides.
Learn More
Offers one or more environmental benefits itemized in the U.S. FTC Green Guides.
Learn More
The BioTek 800 TS absorbance reader is an affordable, high-quality microplate reader for assays in 6- to 384-well formats. For Research Use Only. Not for use in diagnostic procedures.
| Detector Type | Photodiode |
|---|---|
| Reading Options | Endpoint, Kinetic, Well-area Scanning |
| Shaking | Linear (except NB configurations) |
| Product Type | Microplate Reader |
| Voltage | 100/240 V |
| Bandwidth | 10 nm |
| Software Type | Gen5 Data Analysis Software, Gen5 Secure for 21 CFR Part 11 compliance (option) |
| Weight (English) | 18.5 lb. |
| Certifications/Compliance | CE and TUV marked. RoHS compliant. |
| For Use With (Application) | Measuring absorbance in microplate-based assays |
| Light Source | Tungsten Halogen Lamp |
| Interface | USB Type B, USB Type A |
| Weight (Metric) | 8 kg |
Agilent BioTek 406 FX Washer Dispenser
The 406 FX is a compact instrument offering fast, full plate washing along with up to six reagent dispensers. For Research Use Only.
Agilent BioTek Synergy LX Multi-Mode Reader, IVD
Greener Choice
Offers one or more environmental benefits itemized in the U.S. FTC Green Guides.
Learn More
Offers one or more environmental benefits itemized in the U.S. FTC Green Guides.
Learn More
The BioTek Synergy LX multi-mode reader automates many common microplate assays. For In Vitro Diagnostic Use.
| Width (English) | 15 in. |
|---|---|
| Shaking | Linear, Orbital, Double Orbital |
| Software Type | Full data analysis and reporting (under Gen5 control) |
| Absorbance Range | 200 to 900 nm |
| Light Source | Xenon Flash |
| For Use With (Application) | Nucleic acid and protein quantification, ELISA, BCA, Bradford and cell viability |
| Width (Metric) | 38.1 cm |
| USB Connections | 2 |
| Detector Type | Photodiode,PMT |
| Reading Options | End Point,Kinetic,Area Scanning,Absorbance Spectral Scanning |
| Filters | Monochromator Bandpass |
| Accuracy | Wavelength: ±2 nm; Optical Density: <1% at 2.0 OD, <3% at 2.5 OD |
| Voltage | 100/240 V |
| Bandwidth | ≤5 nm |
| Certifications/Compliance | CE and TUV marked. RoHS compliant. |
| Weight (Metric) | 12.3 kg |
| Warranty | 1 year |
| Depth (English) | 15 in. |
| Software Language Versions | English |
| Sensitivity | Fluorescence Intensity: Fluorescein 2 pM; Luminesence: 10 amol ATP |
| Linearity | <1% from 0 to 2.5 OD |
| Depth (Metric) | 38.1 cm |
| Measurement Speed | 24 sec. (96 wells), 76 sec. (384 wells) |
| Frequency | 50/60 Hz |
| Weight (English) | 27 lb. |
| Power Consumption | 24 VDC power supply compatible with 100/240 VAC at 50/60 Hz, 60 W (max.) |
| Resolution | 0.001 OD |
Agilent BioTek Epoch Microplate Spectrophotometer, IVD
Greener Choice
Offers one or more environmental benefits itemized in the U.S. FTC Green Guides.
Learn More
Offers one or more environmental benefits itemized in the U.S. FTC Green Guides.
Learn More
Epoch microplate spectrophotometer has monochromator-based optics, for a filter-free, wide wavelength range for UV-Vis absorbance measurements. For In Vitro Diagnostic Use.
| Height (Metric) | 19.6 cm |
|---|---|
| Cuvette Size | 0.5 mm |
| Width (English) | 12 in. |
| Software Type | Gen5 Data Analysis Software |
| Absorbance Range | 200 to 999 nm, 1 nm increments |
| Light Source | Xenon Flash Lamp |
| Width (Metric) | 30.5 cm |
| Accuracy | ±2 nm |
| Precision | ±0.2 nm |
| Product Type | Microplate Reader |
| Voltage | 100/240 V |
| Bandwidth | 5 nm |
| Weight (English) | <15 lb. |
| Certifications/Compliance | CE and TUV marked. RoHS compliant. |
| Hertz | 50/60 Hz |
| Wavelength Range | 200 to 999 nm |
| Weight (Metric) | 6.8 kg |
| Resolution | 0.0001 OD |
| Height (English) | 7.7 in. |
| Plate Type | 6 to 384-well Plates |
Agilent BioTek MicroFill 96-/384-Well Microplate Dispenser
Greener Choice
Offers one or more environmental benefits itemized in the U.S. FTC Green Guides.
Learn More
Offers one or more environmental benefits itemized in the U.S. FTC Green Guides.
Learn More
MicroFill is an economical, compact, and reliable dispenser for 24-, 96-, and 384-well microplates.
Agilent BioTek Epoch Microplate Spectrophotometer
Greener Choice
Offers one or more environmental benefits itemized in the U.S. FTC Green Guides.
Learn More
Offers one or more environmental benefits itemized in the U.S. FTC Green Guides.
Learn More
Epoch microplate spectrophotometer has monochromator-based optics, for a filter-free, wide wavelength range for UV-Vis absorbance measurements. For Research Use Only. Not for use in diagnostic procedures.
| Height (Metric) | 19.6 cm |
|---|---|
| Cuvette Size | 0.5 mm |
| Width (English) | 12 in. |
| Absorbance Range | 200 to 999 nm, 1 nm increments |
| Light Source | Xenon Flash Lamp |
| Width (Metric) | 30.5 cm |
| Accuracy | ±2 nm |
| Precision | ±0.2 nm |
| Product Type | Microplate Reader |
| Voltage | 100/240 V |
| Bandwidth | 5 nm |
| Weight (English) | <15 lb. |
| Certifications/Compliance | CE and TUV marked. RoHS compliant. |
| Hertz | 50/60 Hz |
| Wavelength Range | 200 to 999 nm |
| Weight (Metric) | 6.8 kg |
| Resolution | 0.0001 OD |
| Height (English) | 7.7 in. |
| Plate Type | 6 to 384-well Plates |
Agilent BioTek Synergy HTX Multi-Mode Microplate Reader, IVD
Greener Choice
Offers one or more environmental benefits itemized in the U.S. FTC Green Guides.
Learn More
Offers one or more environmental benefits itemized in the U.S. FTC Green Guides.
Learn More
The BioTek Synergy HTX is a compact, economical multi-mode reader for 6- to 384-well microplates and Take3 microvolume plates. For In Vitro Diagnostic Use.
| Width (English) | 16 in. |
|---|---|
| Shaking | Linear, Orbital |
| Software Type | Gen5 Microplate Reader and Imager Software,Gen5 Secure for 21 CFR Part 11 compliance (option) |
| Absorbance Range | 200 to 999 nm, 1 nm increments |
| Light Source | Xenon Flash,Tungsten Halogen (Alpha Detection/Fluorescence Intensity) |
| Width (Metric) | 40.6 cm |
| USB Connections | 2 |
| Detector Type | Photodiode,PMT |
| Reading Options | End Point,Kinetic,Spectral Scanning,Well Area Scanning |
| Accuracy | Absorbance: ±2 nm; <1% at 2.0 OD, <3% at 3.0 OD; Dispense: ±1 μL or 2% |
| No. of Dispensers | 2 Syringe Pumps |
| Voltage | 100/240 V |
| Certifications/Compliance | CE and TUV marked. RoHS compliant. |
| Weight (Metric) | 18 kg |
| Height (English) | 10 in. |
| Height (Metric) | 25.4 cm |
| Depth (English) | 15 in. |
| Incubator Temperature | 4-Zone Incubation to 50°C with Condensation Control (±0.2°C at 37°C) |
| Dynamic Range | Absorbance: 0 to 4 OD; Fluorescence Intensity/Luminesence: >6 decades |
| Product Type | Multi-Mode Microplate Reader |
| Depth (Metric) | 38 cm |
| Measurement Speed | 31 sec. (96 wells), 80 sec. (384 wells) |
| Frequency | 50/60 Hz |
| Weight (English) | 40 lb. |
| Compatibility | BioStack and Third Party Automation Compatible |
| Plate Type | 6- to 384-well Plates |
Agilent BioTek Cytation C10 Confocal Imaging Reader
The BioTek Cytation C10 confocal imaging reader combines automated confocal and widefield microscopy with conventional multi-mode microplate reading. For Research Use Only. Not for use in diagnostic procedures.
| Electrical Requirements | Instrument external 250 W, 24 VDC power supply |
|---|---|
| Warranty | 1 year |
| Width (English) | 27 in. |
| Shaking | Linear, Orbital, Double Orbital |
| Absorbance Range | 230 to 999 nm |
| Light Source | Xenon Flash |
| Width (Metric) | 68.6 cm |
| Dynamic Range | Absorbance: 0 to 4 OD; Fluorescence: 7 decades; Luminescence: >6 decades |
| Detection Method | UV-Vis absorbance, Fluorescence intensity, Luminescence |
| Accuracy | <1% (at 3 OD) |
| No. of Dispensers | 2 Syringe Pumps (optional) |
| Dispense Volume | 5 to 1000 μL, 1 μL increments (optional with injectors) |
| Voltage | 100/240 V |
| Bandwidth | 4 nm (230 to 285 nm), 8 nm (>285 nm) |
| Frequency | 50/60 Hz |
| Weight (English) | 122 lb. |
| Software | Gen5 Microplate Reader and Imager Software included; Gen5 Secure for 21 CFR Part 11 compliance (option); Gen5 Image+ and Image Prime software available for full image analysis (option) |
| Certifications/Compliance | CE and TUV marked. RoHS compliant. |
| Temperature Range (Metric) | 4-Zone incubation to 45°C with Condensation Control |
| O2 Concentration Range | 1 to 19% with optional gas controller |
| CO2 Concentration Range | 0 to 20% |
| Weight (Metric) | 53.3 kg |
| Plate Type | Monochromator: 6- to 384-well plates; Imaging: 6- to 1536-well plates |
Agilent BioTek Cytation 7 Cell Imaging Multi-Mode Reader
Greener Choice
Offers one or more environmental benefits itemized in the U.S. FTC Green Guides.
Learn More
Offers one or more environmental benefits itemized in the U.S. FTC Green Guides.
Learn More
BioTek Cytation 7 cell imaging multi-mode reader combines automated digital upright and inverted widefield microscopy with conventional multi-mode microplate reading. For Research Use Only. Not for use in diagnostic procedures.
| Scan Speed | 96-wells: 11 s; 384-wells: 22 s |
|---|---|
| Shaking | Linear, Orbital, Double Orbital |
| Light Source | Xenon Flash |
| Wavelength | Luminescence 300 to 700 nm; Absorbance 230 to 999 nm, 1 nm increment |
| Detection Method | Monochromator- based fluorescence intensity, luminescence and absorbance |
| Reading Options | End point, kinetic, spectral scanning and well area scanning |
| Sensitivity | Fluorescein 2.5 pM (0.25 fmol/well, 384-well plate) – top Fluorescein 4 pM (0.4 fmol/well, 384-well plate) – bottom; luminescence 20 amol ATP (flash) |
| Type | Automated Microscope/Multi-Mode Microplate Reader |
| No. of Dispensers | 2 Syringe Pumps |
| Dispense Volume | 5 to 1000 μL, 1 μL increments |
| Linearity | <1% from 0 to 3 OD |
| Voltage | 100/240 V |
| Bandwidth | 4 nm (230 to 285 nm), 8 nm (>285 nm) |
| Frequency | 50/60 Hz |
| Certifications/Compliance | CE and TUV marked. RoHS compliant. |
| Wavelength Selection | Quad monochromators (top/bottom) |
| Temperature Range (Metric) | 4-Zone incubation to 45°C with Condensation Control (±0.5°C at 37°C) |
| Resolution | 0.0001 OD |
| Plate Type | Monochromator: 6- to 384-well plates; Imaging: 6- to 1536-well plates |
Agilent BioTek Epoch 2 Microplate Spectrophotometer, IVD
Greener Choice
Offers one or more environmental benefits itemized in the U.S. FTC Green Guides.
Learn More
Offers one or more environmental benefits itemized in the U.S. FTC Green Guides.
Learn More
The BioTek Epoch 2 microplate spectrophotometer delivers excellent performance for UV-Vis absorbance measurements. For In Vitro Diagnostic Use.
| Detector Type | Photodiode |
|---|---|
| Accuracy | ±2 nm |
| Measurement Speed | 96 wells: 8 sec., 384 wells: 14 sec. |
| Certifications/Compliance | CE and TUV marked. RoHS compliant. |
| For Use With (Application) | Nucleic acid and protein quantification to ELISA and microbial growth assays |
| Light Source | Xenon Flash Lamp |
| Hertz | 50/60 Hz |
| Wavelength Range | 200 to 999 nm, 1 nm increments |
| Height (English) | 22 in. |
Agilent BioTek 405LS Microplate Washer
Greener Choice
Offers one or more environmental benefits itemized in the U.S. FTC Green Guides.
Learn More
Offers one or more environmental benefits itemized in the U.S. FTC Green Guides.
Learn More
The BioTek 405 LS washer has a well earned reputation as an economical plate washer that offers excellent performance and reliability.
| Height (Metric) | 25.4 cm |
|---|---|
| Depth (English) | 17 in. |
| Width (English) | 14 in. |
| Width (Metric) | 35.6 cm |
| Flow Rate | High to Low Flow; Optimized rates for cell assays |
| Type | Microplate Washer |
| Bottles | Optional (4 or 10 L) |
| Washing Speed | 96-wells: 300 μL/well, 3 cycles: <30 s; 384-wells: 100 μL/well, 3 cycles: <80 s; 384-wells: 400 μL/well, 1 cycle: <20 s |
| Dispense Volume | 25 to 3000 μL/well |
| Precision | <3% CV: 300 μL/well (96-well washing); <4% CV: 80 μL/well (384-well washing) |
| Depth (Metric) | 43.2 cm |
| Wash Cycles | 1 to 250 |
| Voltage | 100/240 V |
| Humidity | 10 to 85%, noncondensing |
| Soak Time | Programmable in minutes and seconds, up to 60 min. |
| Increments | 1 μL |
| Frequency | 50/60 Hz |
| Residual Volume | <2 μL/well (96- and 384-well plates), 96-tube manifold for 96 wells, 192-tube for 384 wells |
| Weight (English) | With internal buffer switching - 36 lb.; Without - 30 lb. |
| Certifications/Compliance | CE and TUV marked. RoHS compliant. |
| Height (English) | 10 in. |
| Weight (Metric) | With internal buffer switching - 16.5 kg; Without - 13.5 kg |
| Plate Type | 96- and 384-well, Low Profile and Standard, Solid and Filter Bottom (Pore Sizes 0.45 to 1.2 μm) Plates |
Agilent BioTek Cytation 5 Cell Imaging Multi-Mode Reader
Greener Choice
Offers one or more environmental benefits itemized in the U.S. FTC Green Guides.
Learn More
Offers one or more environmental benefits itemized in the U.S. FTC Green Guides.
Learn More
Cytation 5 combines automated microscopy and conventional microplate detection in a configurable, upgradable platform. For Research Use Only. Not for use in diagnostic procedures.
| Height (Metric) | 44.5 cm |
|---|---|
| Depth (English) | 20.2 in. |
| Width (English) | 16.4 in. |
| Contrast Methods | Fluorescence, phase contrast, brightfield, high contrast brightfield, color brightfield and phase contrast |
| Shaking | Linear, Orbital, Double Orbital |
| Wattage | 250 W max. |
| Objectives | 1.25x, 2.5x (2.25x eff), 2.5x (2.75x eff), 4x, 10x, 20x, 40x, 60x |
| Absorbance Range | 230 to 999 nm, 1 nm increments |
| Light Source | Xenon Flash; 100 mW 680 nm laser (Alpha Detection) |
| Width (Metric) | 41.6 cm |
| Dynamic Range | Absorbance: 0 to 4 OD; Fluorescence Intensity: 7 decades; Luminesence: >6 decades |
| Reading Options | End point, kinetic, spectral scanning and well area scanning |
| Accuracy | Monochromator: ±2 nm; Dispenser: ±1 μL or 2% |
| Type | Cell Imaging Multi-Mode Microplate Reader |
| No. of Dispensers | 2 Syringe Pumps |
| Dispense Volume | 5 to 1000 μL, 1 μL increments |
| For Use With (Equipment) | BioStack and Third Party Automation Compatible; BioSpa 8 Automated Incubator Compatible |
| Depth (Metric) | 51.4 cm |
| Weight (English) | 80 lb. |
| Certifications/Compliance | CE and TUV marked. RoHS compliant. |
| Camera | Sony CMOS, 16-bit Gray Scale, Standard or WFOV |
| Temperature Range (Metric) | 4-Zone incubation to 65°C with Condensation Control (±0.2°C at 37°C) |
| CO2 Concentration Range | 0 to 20% |
| Weight (Metric) | 36.3 kg |
| Height (English) | 17.5 in. |
Agilent BioTek EL406 Microplate Washer/Dispenser, IVD
Greener Choice
Offers one or more environmental benefits itemized in the U.S. FTC Green Guides.
Learn More
Offers one or more environmental benefits itemized in the U.S. FTC Green Guides.
Learn More
The BioTek EL406 washer dispenser is a compact instrument offering fast, full plate washing along with three reagent dispensers. For In Vitro Diagnostic Use.
| Height (Metric) | 31.8 cm |
|---|---|
| Depth (English) | 18 in. |
| Width (English) | 16.5 in. |
| Width (Metric) | 42 cm |
| Flow Rate | High to Low Flow; Optimized rates for cell assays |
| Type | Microplate Washer/Dispenser |
| Accuracy | ±3%; Dispensing: ±1 μL at 5 μL, +1 μL at 20 μL, ±1% at 100 μL |
| Bottles | Optional (1 or 2 L) |
| Dispense Volume | 500 nL to 3000 μL/well |
| Precision | Wash: <3% CV (model dependent); Dispense (Syringe Pump): ≤5% CV at 5 μL, ≤2.5% CV at 20 μL, ≤1% CV at 100 μL |
| Depth (Metric) | 46 cm |
| Wash Cycles | 1 to 250 |
| Voltage | 100/240 V |
| Soak Time | Programmable in minutes and seconds, up to 60 min. |
| Increments | 1 μL |
| Frequency | 50/60 Hz |
| Residual Volume | ≤2 μL/well |
| Weight (English) | 32 lb. |
| Certifications/Compliance | CE and TUV marked. RoHS compliant. |
| Height (English) | 12.5 in. |
| Weight (Metric) | 14.5 kg |
| Plate Type | 96-, 384- and 1536-well; Low profile and standard height; Solid and filter bottom (option) plates |