DuPont Tyvek IsoClean Single-Use Garments
- Overview
- Coveralls
- Shoe Covers
- Sleeves
The DuPont™ Controlled Environments portfolio offers a comprehensive selection of single-use cleanroom garments and accessories designed for use in pharmaceutical, medical device, and biotech facilities that require high standards for microbiological contamination control. They are comfortable, durable, and available for different cleanroom applications.
- Quality – Single-use apparel provides consistent and reliable quality and eliminates concerns about cross-contamination that can occur with laundering or compromised performance due to multiple wash and sterilization cycles
- Flexibility – The DuPont single-use apparel program allows you to order only the quantities that you plan to use, which offers flexibility as your needs change
- Cost Control – Single-use garments eliminate budget uncertainties associated with garment repair, damage, and loss, helping you to better predict expenditures
Safety Apparel
Choose from a wide selection of DuPont coveralls and suits for use in cleanrooms, production facilities, research labs, and more.
Bulk Coveralls
Features:
- HDPE
- Class 100
- High-density polyethylene material
- Serged seam style
- Binding front zipper
- Elastic wrists and ankles
- Anti-static
- Recyclable
- Low linting
Sterile Coveralls with Dolman Sleeves
Features:
- Dolman sleeves
- Bound seam
- Bound neck
- Zipper entry
- Covered elastic wrists and ankles
- Sterile
Collared Coveralls
Features:
- Standard collar
- Tychem™ IsoClean™
- Tyvek™ flashspun polyethylene
- Serged seams
- Elastic wrist and ankle
- Zipper closure
- Set sleeve
Bulk Packed Shoe Covers
Features:
- Serged seams
- Elastic openings
- Elastic toes
- Bulk packed
- PVC sole
Bulk Packed Sterile Sleeves
Features:
- Tyvek™ IsoClean™
- Tyvek™ flashspun polyethylene
- Bound seams and covered elastic wrists
- Bicep measures 18 inches in length
- Individually packaged
- Sterilized by gamma irradiation