UserName
Viewing profile as user:
Fisher Scientific TraceableLIVE Thermometers
Fisher Scientific TraceableLIVE Thermometers
Fisher Scientific™ TraceableLIVE™ Wi-Fi Datalogging Refrigerator/Freezer Thermometers with Remote Notification
With this new product, you can stay connected to your critical environments with your smartphone, tablet or computer. TraceableLIVE is the simple, efficient and reliable way to experience wireless technology in the lab.
With TraceableLIVE you can:
  • Set device alarm parameters remotely through mobile app or web interface
  • View current conditions in your lab anywhere
  • Generate and email data reports to unlimited number of users
  • Assign security levels and location-specific device access to users
Traceable to NIST for Accuracy
Multi-point calibration and a range of -50°C to 60°C; Accuracy: 0.25°C; Resolution: 0.01°, on an individually numbered Traceable™ Certificate which ensures accuracy from an ISO/IEC 17025:2005 (1750.01) calibration laboratory accredited by A2LA. It indicates traceability to standards provided by NIST (National Institute of Standards and Technology). Requires TraceableLIVE subscription.