UserName
Viewing profile as user:
BD Horizon Real Dye Reagent Guide
BD Horizon Reagent Guide
Minimize Spillover. Maximize Breakthroughs.
BD Horizon™ RealViolet™, RealBlue™, RealYellow™, and RealRed™ Fluorochromes are engineered to minimize cross-laser spillover and enhance panel resolution, enabling high-quality data and flexible panel design across both conventional and spectral cytometers.
BD Horizon Real Fluorochromes
Switch to BD Horizon™ Real Fluorochromes to enjoy the following:
Reduced spillover: Lower cross-laser excitation allows for cleaner emission profiles, which simplifies panel design and unlocks greater flexibility, giving you high-quality panels.
Excellent resolution of diverse antigen densities: These dyes range from moderate to very bright, giving you the ability to assign fluorochromes where they fit best.
Robust intracellular performance: BD Horizon™ Real Fluorochromes enable analysis of intracellular proteins like cytokines, transcription factors and phosphorylated signaling molecules, enabling flexibility in various protocols.
Lot-to-lot consistency and stability: These fluorochromes help provide lot-to-lot consistency across made-to-stock and BD OptiBuild™ Reagents, with photostability under typical lab lighting, so your results stay accurate and reproducible.
Becton Dickinson logo
Cleaner Fluorochromes Are Here
Cleaner Fluorochromes Are Here
Swap out your PerCP and PE tandem dyes with BD Horizon™ Real Dyes
Discover Brighter Alternatives
BD Horizon RealRed Reagents
BD Horizons RealRed™ Reagents
Discover a bright, clean option for spectral flow cytometry panels, excited primarily by the red laser (627-640 nm), and get support for the detection of low expression surface and intracellular markers.
BD Horizon RealYellow Reagents
BD Horizon RealYellow™ Reagents
Spend less time optimizing panels for PE or PE tandem fluorochromes, and opt for BD RealYellow to support the detection of markers with varying levels of expression.
Explore More
For Research Use Only. Not for use in diagnostic or therapeutic procedures
Contact Us
Further your clinical and research applications with BD Horizon Real Dyes reagents. Find the reagents you need for your flow cytometry applications. Achieve robust results and breakthrough discoveries, improve lab efficiency, and help lower costs by reducing the need for repeat experiments.
Fill Out the Form
To learn more and request samples, fill in the information requested, and a Fisher Scientific sales representative will reach out to you.
Required Field *
Company
Address