UserName
Viewing profile as user:
R&D Systems Immunoassays
R&D Systems Immunoassays
Immunoassays You Can Trust
R&D Systems, a Bio-Techne brand, delivers high-performance immunoassays engineered for precision, reproducibility, and reliable results—helping accelerate your biomarker discovery.
With an extensive, rigorously validated assay portfolio, R&D Systems™ products provide highly specific and sensitive protein quantification across a broad range of cytokines, chemokines, and key biomarkers. Assays and reagents are manufactured in‑house under rigorous quality standards designed to ensure lot‑to‑lot consistency and dependable performance.
When you choose an R&D System assay, you also receive comprehensive product documentation and technical support—empowering you with the information you need to trust every step of your workflow.
Select the ELISA kits and assays that best meet your needs.
Select the ELISA kits and assays that best meet your needs.
Assays for Accurate and Reproducible Results
Save time—and avoid the cost of repeated experiments—with assays and kits that are thoroughly tested for validated linearity, spike recovery, and precision. In-house manufacturing helps ensure R&D Systems reagents provide unmatched accuracy and consistency across complex samples—enabling confident biomarker quantification.
From robust dilution linearity to dependable recovery across low, mid, and high analyte concentrations, the assays are engineered to deliver the performance you need for publication-ready data every time.
Explore the complete assay portfolio and find the right assays for your research needs.
Quantikine ELISA Kits | Luminex Assays | DuoSet ELISA Kits | Comparison Chart
Quantikine ELISA Kits
Quantikine ELISA Kits
Explore ready‑to‑use ELISA kits engineered for accurate, highly sensitive, and reproducible quantitative protein detection—featuring fully validated reagents, optimized workflows, and convenient colorimetric detection options for dependable results every time.
 Luminex Assays
Luminex Assays
Browse R&D Systems™ Luminex™ bead-based multiplex assays, designed to provide accurate, reproducible results for up to 50 analytes per sample.
DuoSet ELISA Kits
DuoSet ELISA Kits and Matched Antibody Pairs
Gain full control over your assay design with flexible R&D Systems™ DuoSet™ buffer preparation and plate coating options—supported by complete systems for building custom sandwich immunoassays using rigorously matched antibody pairs for reliable capture and detection.
Comparison Chart
Assay
Benefits
Number of Analytes
Format
Sample Volume
Throughput
Assay Time
Instrument
DuoSet ELISA Kits
Largest Menu, Economical, Flexible
1
Matched Antibody Pair Sandwich Immunoassay
10 to 200 µL
96- or 384-Well Plate
20 hr
Plate Reader
Quantikine ELISA Kits
Most Published, Gold Standard
1
Matched Antibody Pair Sandwich Immunoassay
10 to 200 µL
96-Well Plate
1.5 to 5 hr
Plate Reader
QuantiGlo ELISA
Broad Dynamic Range
1
Matched Antibody Pair Sandwich Immunoassay with Chemiluminescent Detection
10 to 200 µL
96-Well Plate
4 to 4.5 hr
Luminometer
Quantikine HS ELISA
Highest Sensitivity
1
Matched Antibody Pair Sandwich Immunoassay
10 to 200 µL
96-Well Plate
4 to 4.5 hr
Plate Reader
Quantikine QuicKit ELISA
Fastest Plate-Based ELISA
1
Matched Antibody Pair Sandwich Immunoassay
50 µL
96-Well Plate
90 min
Plate Reader
Proteome Profiler Arrays
Largest Screening Capabilities
Up to 119
Membrane Array
50 to 500 µL
1 sample per membrane
20 hr
Imaging System
Luminex Discovery Assays
Large Multiplex Panels, Customization
Up to 50
Bead-Based (Multiplex Microspheres)
25 to 50 µl
96- or 384-Well Plate
4 hr
Luminex System
Luminex Premixed High-Performance Assays
10 Large Customizable Multiplex Panels, Including 2 High-Sensitivity Panels
Up to 50
Bead-Based (Multiplex Microspheres)
25 to 50 µl
96- or 384-Well Plate
4 hr
Luminex System
Luminex User-Mixed High-Performance Assays
13 Large Customizable Multiplex Panels, Including 2 High-Sensitivity Panels
Up to 50
Bead-Based (Multiplex Microspheres)
25 to 50 µl
96- or 384-Well Plate
4 hr
Luminex System
Luminex Fixed High-Performance Assays
9 Fixed, Large Multiplex Panels, High Sensitivity
Up to 50
Bead-Based (Multiplex Microspheres)
25 to 50 µl
96- or 384-Well Plate
4 hr
Luminex System
Enable Flexibility and Efficiency
Advance your biomarker discovery with over 50 years of R&D Systems assay expertise and industry leading immunoassay performance.
From routine studies to complex clinical research, R&D Systems offers one of the largest portfolios of immunoassays—supporting everything from singleplex ELISAs to highplex R&D Systems™ Luminex™ panels. Rely on rigorously validated matched antibody pairs and fully customizable Luminex panels to tailor your assay design to your research needs.
Accelerate your workflow with innovative, time-saving solutions like R&D Systems™ Quantikine™ QuicKit™ ELISAs, delivering trusted Quantikine performance in just 90 minutes. For flexible, cost effective development, R&D Systems™ DuoSet™ ELISA Development Systems provide affordable assay build up and scale-up—reducing screening time, optimizing antibody pair selection, and minimizing repeat experiment costs.
Need help selecting the right immunoassays? Fill out the form and let Fisher Scientific specialists guide you to the best R&D Systems solutions for your biomarker discovery goals.
Reach Out to a Representative
Complete the form to request to be contacted by a representative about R&D Systems immunoassays.
Required Field *
Company
Address
Country
State