Thermo Scientific SurePac MDi HPLC Columns
Maximize Efficiency, Minimize Interactions
Thermo Scientific™ SurePac™ Bio 550 SEC MDi™ HPLC analytical columns and guards let you experience the future of size-exclusion chromatography (SEC) with their design that delivers robust and reproducible performance at high resolution and throughput to accelerate your therapeutics analysis process.
Achieve Next-Level Separation with Groundbreaking Technology
Count on high-performance SurePac Bio 550 SEC MDi Columns to elevate your separation. Developed using groundbreaking MDi technology, they integrate an advanced monodisperse silica particle platform with biocompatible, inert hardware for executing reliable size-exclusion chromatography.
- Leverage the power of 3 μm monodisperse silica particles for robust, reproducible, efficient separations and high throughput, outperforming traditional polydisperse 5 μm media
- Bioinert hardware eliminates secondary interactions and optimizes sample recovery
- Wide pores (550 Å) enable the separation of aggregates in adeno-AAVs and large proteins, and the size-based separation of nucleic acids
Featured Products
Catalog Number 03-452-330
Catalog Number 03-452-269