Filtered Search Results
Ansell MICROFLEX™ UltraSense™ US-220 Nitrile Gloves
Greener Choice
Offers one or more environmental benefits itemized in the U.S. FTC Green Guides.
Learn More
Offers one or more environmental benefits itemized in the U.S. FTC Green Guides.
Learn More
Powder free, comfortable nitrile examination gloves offer durability when worn for extended periods of time. Ideal for healthcare, life science and food processing applications.
| Tensile Strength | 30 MPa |
|---|---|
| Disposable | Single-use |
| Length (Metric) | 24.38 cm |
| Applications | Industrial,Healthcare |
| Chemotherapeutic Rated | No |
| Finger Thickness (English) | 4.3 mil |
| Color | Blue |
| Length (English) | 9.6 in. |
| Material | Nitrile |
| Powdered | Powder Free |
| Finger Thickness (Metric) | 0.11 mm |
| Thickness (Metric) | 0.07 mm |
| Series | MICROFLEX™ UltraSense™ US-220 |
| FDA 510 (k) | Yes |
| Certifications/Compliance (Europe) | EN 374:2003, EN 420:2003 + A1:2009 |
| Thickness (English) | 3.1 mil |
| Palm Thickness (English) | 3.1 mil |
| Latex Free | Yes |
| Exterior Finish | Partially Textured |
| Certifications/Compliance | ASTM D6319, FDA21 CFR 177-2600 |
| AQL | 1.5 |
| Palm Thickness (Metric) | 0.08 mm |
Ansell MICROFLEX™ UltraSense™ EC USE-880 Nitrile Gloves
Greener Choice
Offers one or more environmental benefits itemized in the U.S. FTC Green Guides.
Learn More
Offers one or more environmental benefits itemized in the U.S. FTC Green Guides.
Learn More
Extended cuff offers extra protection over the wrist and forearm for use in high-risk environments.
| Tensile Strength | 33 MPa |
|---|---|
| Disposable | Single-use |
| Length (Metric) | 29 cm |
| Applications | Healthcare,Laboratory |
| Chemotherapeutic Rated | No |
| Finger Thickness (English) | 5.1 mil |
| Color | Blue |
| Length (English) | 11.4 in. |
| Material | Nitrile |
| Powdered | Powder Free |
| Finger Thickness (Metric) | 0.13 mm |
| Thickness (Metric) | 0.13 mm |
| Series | UltraSense™ EC USE-880 |
| FDA 510 (k) | Yes |
| Certifications/Compliance (Europe) | EN 420:2003 + A1:2009 |
| Thickness (English) | 5.1 mil |
| Palm Thickness (English) | 3.9 mil |
| Latex Free | Yes |
| Exterior Finish | Partially Textured |
| Certifications/Compliance | ASTM D 6319 |
| Palm Thickness (Metric) | 0.10 mm |
KleenGuard™ Nemesis™ Eyewear
Greener Choice
Offers one or more environmental benefits itemized in the U.S. FTC Green Guides.
Learn More
Offers one or more environmental benefits itemized in the U.S. FTC Green Guides.
Learn More
KleenGuard™ Nemesis™ Safety Eyewear is stylish, durable and have a lightweight construction.
| Disposable | Reusable |
|---|---|
| Includes | Neck Cord |
| Frame Color | Black |
| Lens Material | Plastic |
| Product Type | Safety Glasses |
| Material | Plastic |
| Certifications/Compliance | Meets ANSI/ISEA Z87.1+ standards. |
Ansell MICROFLEX™ Integra™ N86 Nitrile Gloves
Integra™ nitrile examination gloves are designed with extra thickness in the palm and finger to offer added durability.
| Tensile Strength | 31 MPa |
|---|---|
| Disposable | Yes |
| Length (Metric) | 24.13 cm |
| Applications | Industrial,Healthcare,Laboratory |
| Chemotherapeutic Rated | No |
| Finger Thickness (English) | 7.5 mil |
| Color | Blue |
| Length (English) | 9.5 in. |
| Material | Nitrile |
| Powdered | Powder Free |
| Finger Thickness (Metric) | 0.19 mm |
| Thickness (Metric) | 0.17 mm |
| Series | MICROFLEX™ Integra™ N86 |
| FDA 510 (k) | Yes |
| Certifications/Compliance (Europe) | EN 420:2003 + A1:2009 |
| Thickness (English) | 6.7 mil |
| Palm Thickness (English) | 6.7 mil |
| Latex Free | Yes |
| Exterior Finish | Fully Textured |
| Certifications/Compliance | ASTM D6319, FDA21 CFR 177-2600, ISO 9001 |
| AQL | 1.5 |
| Palm Thickness (Metric) | 0.17 mm |
Ansell MICROFLEX™ V28 Industrial Vinyl Gloves
Vinyl gloves with a smooth finish are ideal for food preparation, light-duty cleaning and general maintenance applications.
| Length (Metric) | 24 cm |
|---|---|
| Applications | Industrial |
| Finger Thickness (English) | 5.1 mil |
| Color | Clear |
| Length (English) | 9.5 in. |
| Material | Vinyl |
| Powdered | Powder Free |
| Finger Thickness (Metric) | 0.13 mm |
| Thickness (Metric) | 0.13 mm |
| Series | MICROFLEX™ V28 |
| Certifications/Compliance (Europe) | EN 420:2003 + A1:2009 |
| Thickness (English) | 5.1 mil |
| Palm Thickness (English) | 2.8 mil |
| Exterior Finish | No Texture |
| Certifications/Compliance | FDA 21 CFR 177-2600 |
| AQL | 4 |
| Palm Thickness (Metric) | 0.07 mm |
Ansell MICROFLEX™ CE5-512 Clean Latex Gloves
Comfortable latex gloves are designed for use in Class 100/ISO 5 Cleanroom environments.
| Length (Metric) | 30.5 cm |
|---|---|
| Finger Thickness (English) | 7.1 mil |
| Color | Natural |
| Length (English) | 12 in. |
| Material | Latex |
| Powdered | Powder Free |
| Sterility | Non-sterile |
| Hand | Ambidextrous |
| Finger Thickness (Metric) | 0.18 mm |
| Thickness (Metric) | 0.18 mm |
| Series | MICROFLEX™ CE5-512 |
| Certifications/Compliance (Europe) | Category III, EN 420:2003 + A1:2009, EN 455-1 |
| Thickness (English) | 7.1 mil |
| Palm Thickness (English) | 3.3 mil |
| Cleanroom Class | Class 100 (ISO 5) |
| Latex Free | No |
| Exterior Finish | Partially Textured |
| Certifications/Compliance | ASTM D3767,ASTM D3577,D412-06a |
| Manufacturing/QMS Audit Standards | ISO 9001:2008 |
| AQL | 1.5 |
| Palm Thickness (Metric) | 0.14 mm |
Ansell MICROFLEX™ Soft White Nitrile TQ-601 Gloves
Greener Choice
Offers one or more environmental benefits itemized in the U.S. FTC Green Guides.
Learn More
Offers one or more environmental benefits itemized in the U.S. FTC Green Guides.
Learn More
Thin, powder free gloves with HYDRASOFT™ comfortably protects hands while replenishing moisture and preventing dryness.
| Tensile Strength | 24 MPa |
|---|---|
| Disposable | Yes |
| Length (Metric) | 24.38 cm |
| Applications | Healthcare,Industrial |
| Chemotherapeutic Rated | No |
| Finger Thickness (English) | 3.9 mil |
| Color | White |
| Length (English) | 9.6 in. |
| Material | Nitrile |
| Powdered | Powder Free |
| Finger Thickness (Metric) | 0.10 mm |
| Thickness (Metric) | 0.07 mm |
| Series | MICROFLEX™ Soft White Nitrile TQ-601 |
| FDA 510 (k) | Yes |
| Certifications/Compliance (Europe) | EN 420:2003 + A1:2009, EN 455 1-4, EN ISO 13485 |
| Thickness (English) | 2.8 mil |
| Palm Thickness (English) | 2.8 mil |
| Latex Free | Yes |
| Exterior Finish | Partially Textured |
| Certifications/Compliance | ASTM D6319, FDA21 CFR 177-2600 |
| AQL | 1.5 |
| Palm Thickness (Metric) | 0.07 mm |
Ansell X-Tra™ Traction Shoe Covers
Greener Choice
Offers one or more environmental benefits itemized in the U.S. FTC Green Guides.
Learn More
Offers one or more environmental benefits itemized in the U.S. FTC Green Guides.
Learn More
Shoe covers made with SMS fabric feature adhesive traction strips on the bottom for stability.
| Includes | Traction |
|---|---|
| Disposable | Single-use |
| Sole Type | Traction Strips |
| Product Type | Shoe Covers |
| Color | Blue |
| Material | SMS |
| Coating | Polyethylene |
| For Use With (Application) | Laboratories, Healthcare |
| Closure Type | Elastic Ankle |
| Sterility | Non-sterile |
Ansell KleenGuard™ A40 Liquid and Particle Protection Lab Coats
Greener Choice
Offers one or more environmental benefits itemized in the U.S. FTC Green Guides.
Learn More
Offers one or more environmental benefits itemized in the U.S. FTC Green Guides.
Learn More
Microporous lab coats provide protection against debris, non-hazardous liquid chemicals, and dry particulates.
| Product Type | General Purpose Lab Coat |
|---|---|
| Color | White |
| Style | Knee-length |
| Material | Microporous Film |
| Certifications/Compliance | NFPA 99, ASTM F1670, ANSI ISEA 101-1996 |
| Gender | Unisex |
| For Use With (Application) | Fiberglass Manufacturing, Tank Cleaning, Utilities, Liquid Handling, Pressure Washing, Maintenance, General Manufacturing |
| Closure Type | Snaps |
| Collar Type | Traditional |
| Sterility | Non-sterile |
Ansell KleenGuard™ A40 Liquid and Particle Protection Shoe Covers
Greener Choice
Offers one or more environmental benefits itemized in the U.S. FTC Green Guides.
Learn More
Offers one or more environmental benefits itemized in the U.S. FTC Green Guides.
Learn More
A40 shoe covers are made with breathable, microporous film to protect from debris, non-hazardous liquid chemicals, and dry particulates.
| Resistance | Fluid |
|---|---|
| Disposable | Single-use |
| Seam Type | Serged |
| Product Type | Shoe Covers |
| Color | White |
| Material | Microporous Fabric |
| Certifications/Compliance | NFPA 99 |
| Coating | Microporous Film Laminate |
| For Use With (Application) | Manufacturing, Painting, Cleaning, Liquid Handling, Utilities, Maintenance |
| Sterility | Non-sterile |
Ansell KleenGuard™ V60 30-06™ Readers Safety Eyewear
Greener Choice
Offers one or more environmental benefits itemized in the U.S. FTC Green Guides.
Learn More
Offers one or more environmental benefits itemized in the U.S. FTC Green Guides.
Learn More
KleenGuard V60 30-06 Readers Safety Glasses are equipped to suit those who need vision assistance and eye protection. Available in a variety of lens and frame colors and prescription strengths, they are designed for long-term comfort.
| Frame Color | Black |
|---|---|
| Lens Tint | Clear |
| UV Protection | UV400 |
| Lens Material | Polycarbonate |
| Frame Material | Nylon |
| Certifications/Compliance | ANSI Z87.1-2010 |
Ansell TouchNTuff™ DermaShield™ 73-701 Neoprene Cleanroom Gloves
SureTRACE
Supports traceability with guaranteed access to certificates and proactive change notifications.
Learn More
Greener Choice
Offers one or more environmental benefits itemized in the U.S. FTC Green Guides.
Learn More
Supports traceability with guaranteed access to certificates and proactive change notifications.
Learn More
Offers one or more environmental benefits itemized in the U.S. FTC Green Guides.
Learn More
Sterile neoprene (polychloroprene) cleanroom gloves are free of proteins and accelerators. Suitable for use in aseptic and Class 100/ISO 5 cleanroom environments.
| Chlorinated | Yes |
|---|---|
| Length (Metric) | 29.5 cm |
| Chemotherapeutic Rated | Yes |
| Finger Thickness (English) | 8.3 mil |
| Length (English) | 11.6 in. |
| Powdered | Powder Free |
| Finger Thickness (Metric) | 0.19 mm |
| Thickness (English) | 8.3 mil |
| Palm Thickness (English) | 7 mil |
| Exterior Finish | No Texture |
| Certifications/Compliance | 21 CFR 820 Quality System Regulations,ASTM D3577,ASTM D7160,ASTM D6978-05,ISO 10282,ISO 11193,ISO 13485,ISO 14001 |
| Manufacturing/QMS Audit Standards | ISO 9001:2015 |
| AQL | 0.65 |
| Tensile Strength | 17 MPa |
| Disposable | Single-use |
| Color | Green |
| Material | Polychloroprene |
| Sterility | Sterile |
| Hand | Specific |
| Thickness (Metric) | 0.19 mm |
| Accelerator Free | Yes |
| Series | TouchNTuff™ 73-701 |
| Certifications/Compliance (Europe) | CE 0493, EN 420:2003 + A1:2009, EN 421:2010, EN 556-1:2001, EN ISO 374-1:2016, EN ISO 374-5:2016 |
| Product Type | Cleanroom Gloves |
| Cleanroom Class | Class 100 (ISO 5) |
| Model | 73-701 |
| Latex Free | Yes |
| Palm Thickness (Metric) | 0.18 mm |
Ansell AlphaTec™ Solvex™ 37-676 Nitrile Chemical Resistant Gloves
Nitrile gloves deliver optimal results in wet or dry work environments where chemical resistance is crucial.
| Chlorinated | Yes |
|---|---|
| Grip Design | Reversed Lozenge |
| Length (Metric) | 33 cm |
| EN388 Standard | EN 388:4101X, EN 388:4101 |
| Finger Thickness (English) | 0.0169 in. |
| Length (English) | 13 in. |
| For Use With (Application) | Chemical plants and transportation, chemical processing, Oil refining and Petrochemicals, cleaning and de-greasing, exposure to chemicals, Solvents, hazardous liquids, PVC cements or standing Water, janitorial and maintenance work |
| Finger Thickness (Metric) | 0.43 mm |
| EN ISO 374-1 | Type A |
| EN ISO 374 - 5 | Bacteria, Fungi only |
| Thickness (English) | 15 mil |
| Palm Thickness (English) | 0.0169 in. |
| Certifications/Compliance | FDA 21 CFR 177.2600 |
| Liner Material | Cotton Flock |
| Glove Cuff Style | Gauntlet |
| EN 421 | No |
| Applications | Industrial |
| Color | Green |
| Material | Nitrile |
| Thickness (Metric) | 0.38 mm |
| Series | AlphaTec™ Solvex™ 37-676 |
| ANSI Abrasion Rating | ANSI 5 |
| Certifications/Compliance (Europe) | EN 1149 |
| EN ISO 374-1 CHEMICAL LETTER CODES | JKLOPT |
| Product Type | Chemical-resistant Gloves |
| Latex Free | Yes |
| Lining | Lined |
| Palm Thickness (Metric) | 0.43 mm |
Ansell HyFlex™ 74-048 Lightweight Cut Protection Gloves
Lightweight gloves provide a high-level of cut protection and dexterity. Can be used as a liner under another glove.
| Construction | Knit |
|---|---|
| Length (Metric) | 23.5 cm |
| EN388 Standard | EN 388:4X2XF, EN388:454X |
| Color | Gray |
| Length (English) | 9.3 in. |
| Cut Resistance Rating | ANSI A6 |
| Material | Dyneema |
| For Use With (Application) | Food processing |
| Series | HyFlex™ 74-048 |
| ANSI Abrasion Rating | ANSI 3 |
| Certifications/Compliance (Europe) | EN 420:2003 + A1:2009 |
| Product Type | Cut-resistant Gloves |
| Latex Free | Yes |
| Liner Material | Dyneema™ |
| Glove Cuff Style | Extended |
Ansell MICROFLEX™ NeoTouch™ 25-201 Neoprene Gloves, Extended Cuff
Greener Choice
Offers one or more environmental benefits itemized in the U.S. FTC Green Guides.
Learn More
Offers one or more environmental benefits itemized in the U.S. FTC Green Guides.
Learn More
Neoprene gloves with an extended cuff offers expanded chemical splash protection over the wrist and forearm.
| Length (Metric) | 29 cm |
|---|---|
| Applications | Laboratory |
| Finger Thickness (English) | 6.3 mil |
| Color | Green |
| Length (English) | 11.5 in. |
| Material | Neoprene |
| Powdered | Powder Free |
| Finger Thickness (Metric) | 0.16 mm |
| Thickness (Metric) | 0.16 mm |
| Series | MICROFLEX™ NeoTouch™ 25-201 |
| Certifications/Compliance (Europe) | EN 374-1:2016, EN 374-5:2016, EN 420:2003 + A1:2009 |
| Thickness (English) | 6.3 mil |
| Palm Thickness (English) | 5.1 mil |
| Exterior Finish | Partially Textured |
| Certifications/Compliance | ASTM D6977,FDA 21 CFR 177-2600 |
| AQL | 1.5 |
| Palm Thickness (Metric) | 0.13 mm |