UserName
Viewing profile as user:
Learn About Agilent BioTek Imaging and Microscopy Products
Simplify Your Selection Process
Find Agilent BioTek imaging and microscopy solutions for a wide range of applications and budgets. You can simplify your selection process with the following details about Agilent BioTek Lionheart Automated Microscopes and Agilent BioTek Cytation Cell Imaging Multi-Mode Readers.
Each product is compatible with or features:
  • 6- to 1536-well plates
  • Slides, cell culture dishes and flasks, hemocytometers, and chamber slides
  • 1.25x, 2.5x, 4x, 10x, 20x, 40x, and 60x magnifications (air)
  • 60x and 100x magnifications (oil, Lionheart only)
Agilent BioTek Lionheart Automated Microscopes include FX and LX models to meet the needs of your laboratory. Perform inverted widefield imaging with the Lionheart LX or complete kinetic live cell imaging workflows with the Lionheart FX.
Agilent BioTek Cytation Cell Imaging Multi-Mode Readers include C10, 7, 5, and 1 models capable of both imaging and plate reading. The Cytation C10 is ideal for inverted widefield and confocal imaging, the Cytation 7 for inverted and upright widefield imaging, the Cytation 5 for advanced inverted widefield imaging, and the Cytation 1 for basic inverted widefield imaging.
Request More Information
Complete this form to learn more about Agilent BioTek microscopy solutions.
A Fisher Scientific sales representative will be in touch to help you find the product that best meets your needs.
Required Field *
Required Field *
Country
State/Province
By submitting your data, you agree to our Privacy Policy and will receive emails based on your preferences.
© Thermo Fisher Scientific, Inc. All Rights Reserved.
Thermo Fisher Scientific Logo
Compare a Range of Cell Imaging and Microscopy Solutions
Features
Lionheart FX
Lionheart LX
Cytation C10
Cytation 7
Cytation 5
Cytation 1
General
Incubation
To 40°C
To 45°C
To 45°C
To 65°C
To 45°C
Joystick Controller Available
x
x
x
x
x
Multi-Mode Plate Reading  
x
x
x
x
BioStack Compatible  
x
x
x
x
BioSpa Compatible   
x
x
x
Agilent BenchCel Compatible    
x
Imaging
Widefield Fluorescence
x
x
x
x
x
x
Confocal Fluorescence
x
High-Contrast Brightfield
x
x
x
x
x
x
Brightfield
x
x
x
x
x
Color Brightfield
x
x
x
x
x
Phase Contrast
x
x
x
x
Inverted Microscope
x
x
x
x
x
x
Upright Microscope
x