Filtered Search Results
Agilent BioTek 800 TS Absorbance Reader
Greener Choice
Offers one or more environmental benefits itemized in the U.S. FTC Green Guides.
Learn More
Offers one or more environmental benefits itemized in the U.S. FTC Green Guides.
Learn More
The BioTek 800 TS absorbance reader is an affordable, high-quality microplate reader for assays in 6- to 384-well formats. For Research Use Only. Not for use in diagnostic procedures.
Detector Type | Photodiode |
---|---|
Reading Options | Endpoint, Kinetic, Well-area Scanning |
Shaking | Linear (except NB configurations) |
Product Type | Microplate Reader |
Voltage | 100/240 V |
Bandwidth | 10 nm |
Software Type | Gen5 Data Analysis Software, Gen5 Secure for 21 CFR Part 11 compliance (option) |
Weight (English) | 18.5 lb. |
Certifications/Compliance | CE and TUV marked. RoHS compliant. |
For Use With (Application) | Measuring absorbance in microplate-based assays |
Light Source | Tungsten Halogen Lamp |
Interface | USB Type B, USB Type A |
Weight (Metric) | 8 kg |
Agilent BioTek 50 TS Microplate Washer
Greener Choice
Offers one or more environmental benefits itemized in the U.S. FTC Green Guides.
Learn More
Offers one or more environmental benefits itemized in the U.S. FTC Green Guides.
Learn More
The BioTek 50 TS washer offers excellent microplate washing performance. For Research Use Only. Not for use in diagnostic procedures.
Wash Volume | 25 to 3000 μL/well |
---|---|
Connections | USB (1) |
Fluid Delivery Type | One positive displacement syringe drive |
Automation Interface | 4.3 in. Color LCD Touch Screen Display |
Flow Rate | 150 to 1000 μL/well/sec. (manifold dependent) |
Type | Microplate Washer |
Bottles | Optional (2 to 4 L) |
Wash Cycles | 1 to 10 |
Soak Time | Programmable in minutes and seconds, up to 30 min. |
Weight (English) | 22 lb. |
Certifications/Compliance | CE and TUV marked. RoHS compliant. |
Display | 4.3 in. LCD Color Touch Screen |
Weight (Metric) | 9.8 kg |
Plate Type | 24-, 96-, 384-well Plates (96- and 384-well: Standard and Low Profile Plates, Solid and Filter Bottom (Option), 24-well Plates with 4-well Manifold) |
Agilent BioTek Synergy LX Multi-Mode Reader
Greener Choice
Offers one or more environmental benefits itemized in the U.S. FTC Green Guides.
Learn More
Offers one or more environmental benefits itemized in the U.S. FTC Green Guides.
Learn More
The BioTek Synergy LX multi-mode reader automates many common microplate assays. NOTE: Fluorescence filter cubes are required for fluorescence measurements (sold separately). For Research Use Only. Not for use in diagnostic procedures.
Width (English) | 15 in. |
---|---|
Shaking | Linear, Orbital, Double Orbital |
Absorbance Range | 200 to 900 nm |
Light Source | Xenon Flash |
For Use With (Application) | Nucleic acid and protein quantification, ELISA, BCA, Bradford and cell viability |
Width (Metric) | 38.1 cm |
USB Connections | 2 |
Detector Type | Photodiode,PMT |
Reading Options | End Point,Kinetic,Area Scanning,Absorbance Spectral Scanning |
Filters | Monochromator Bandpass |
Accuracy | Wavelength: ±2 nm; Optical Density: <1% at 2.0 OD, <3% at 2.5 OD |
Voltage | 100/240 V |
Bandwidth | ≤5 nm |
Certifications/Compliance | CE and TUV marked. RoHS compliant. |
Weight (Metric) | 12.3 kg |
Warranty | 1 year |
Depth (English) | 15 in. |
Software Language Versions | English |
Sensitivity | Fluorescence Intensity: Fluorescein 2 pM; Luminesence: 10 amol ATP |
Product Type | Multi-mode Microplate Reader |
Linearity | <1% from 0 to 2.5 OD |
Depth (Metric) | 38.1 cm |
Measurement Speed | 24 sec. (96 wells), 76 sec. (384 wells) |
Frequency | 50/60 Hz |
Weight (English) | 27 lb. |
Power Consumption | 24 VDC power supply compatible with 100/240 VAC at 50/60 Hz, 60 W (max.) |
Resolution | 0.001 OD |
Agilent BioTek 405LS Microplate Washer
Greener Choice
Offers one or more environmental benefits itemized in the U.S. FTC Green Guides.
Learn More
Offers one or more environmental benefits itemized in the U.S. FTC Green Guides.
Learn More
The BioTek 405 LS washer has a well earned reputation as an economical plate washer that offers excellent performance and reliability.
Height (Metric) | 25.4 cm |
---|---|
Depth (English) | 17 in. |
Width (English) | 14 in. |
Width (Metric) | 35.6 cm |
Flow Rate | High to Low Flow; Optimized rates for cell assays |
Type | Microplate Washer |
Bottles | Optional (4 or 10 L) |
Washing Speed | 96-wells: 300 μL/well, 3 cycles: <30 s; 384-wells: 100 μL/well, 3 cycles: <80 s; 384-wells: 400 μL/well, 1 cycle: <20 s |
Dispense Volume | 25 to 3000 μL/well |
Precision | <3% CV: 300 μL/well (96-well washing); <4% CV: 80 μL/well (384-well washing) |
Depth (Metric) | 43.2 cm |
Wash Cycles | 1 to 250 |
Voltage | 100/240 V |
Humidity | 10 to 85%, noncondensing |
Soak Time | Programmable in minutes and seconds, up to 60 min. |
Increments | 1 μL |
Frequency | 50/60 Hz |
Residual Volume | <2 μL/well (96- and 384-well plates), 96-tube manifold for 96 wells, 192-tube for 384 wells |
Weight (English) | With internal buffer switching - 36 lb.; Without - 30 lb. |
Certifications/Compliance | CE and TUV marked. RoHS compliant. |
Height (English) | 10 in. |
Weight (Metric) | With internal buffer switching - 16.5 kg; Without - 13.5 kg |
Plate Type | 96- and 384-well, Low Profile and Standard, Solid and Filter Bottom (Pore Sizes 0.45 to 1.2 μm) Plates |
Agilent BioTek EL406 Microplate Washer/Dispenser
Greener Choice
Offers one or more environmental benefits itemized in the U.S. FTC Green Guides.
Learn More
Offers one or more environmental benefits itemized in the U.S. FTC Green Guides.
Learn More
The BioTek EL406 washer dispenser is a compact instrument offering fast, full plate washing along with three reagent dispensers.
Depth (English) | 18 in. |
---|---|
Width (English) | 16.5 in. |
Width (Metric) | 42 cm |
Flow Rate | High to Low Flow; Optimized rates for cell assays |
Type | Microplate Washer/Dispenser |
Accuracy | ±3%; Dispensing: ±1 μL at 5 μL, +1 μL at 20 μL, ±1% at 100 μL |
Bottles | Optional (1 or 2 L) |
Washing Speed | 96 wells: 300 μL/well, 96-tube manifold: 13 s; 384 wells: 100 μL/well, 192-tube manifold: 17 s; 1536 wells: 10 μL/wells, two 32-tube manifolds: 36 s |
Dispense Volume | 500 nL to 3000 μL/well |
Precision | Wash: <3% CV (model dependent); Dispense (Syringe Pump): ≤5% CV at 5 μL, ≤2.5% CV at 20 μL, ≤1% CV at 100 μL |
Depth (Metric) | 46 cm |
Wash Cycles | 1 to 250 |
Voltage | 100/240 V |
Soak Time | Programmable in minutes and seconds, up to 60 min. |
Increments | 1 μL |
Frequency | 50/60 Hz |
Residual Volume | ≤2 μL/well |
Weight (English) | 32 lb. |
Certifications/Compliance | CE and TUV marked. RoHS compliant. |
Weight (Metric) | 14.5 kg |
Plate Type | 96-, 384- and 1536-well; Low profile and standard height; Solid and filter bottom (option) plates |
Agilent BioTek Epoch Microplate Spectrophotometer
Greener Choice
Offers one or more environmental benefits itemized in the U.S. FTC Green Guides.
Learn More
Offers one or more environmental benefits itemized in the U.S. FTC Green Guides.
Learn More
Epoch microplate spectrophotometer has monochromator-based optics, for a filter-free, wide wavelength range for UV-Vis absorbance measurements. For Research Use Only. Not for use in diagnostic procedures.
Height (Metric) | 19.6 cm |
---|---|
Cuvette Size | 0.5 mm |
Width (English) | 12 in. |
Absorbance Range | 200 to 999 nm, 1 nm increments |
Light Source | Xenon Flash Lamp |
Width (Metric) | 30.5 cm |
Accuracy | ±2 nm |
Precision | ±0.2 nm |
Product Type | Microplate Reader |
Voltage | 100/240 V |
Bandwidth | 5 nm |
Weight (English) | <15 lb. |
Certifications/Compliance | CE and TUV marked. RoHS compliant. |
Hertz | 50/60 Hz |
Wavelength Range | 200 to 999 nm |
Weight (Metric) | 6.8 kg |
Resolution | 0.0001 OD |
Height (English) | 7.7 in. |
Plate Type | 6 to 384-well Plates |
Agilent BioTek MultiFlo FX Multi-Mode Dispenser PROMO
MultiFlo FX is an automated multi-mode reagent dispenser for 6- to 1536-well microplates.
Height (Metric) | 20.3 cm |
---|---|
Speed | 96 wells, 5 μL cass, 10 μL/well: 3 sec., 96 wells, 5 μL cass, 20 μL/well: 3.5 sec., 384 wells, 5 μL cass, 5 μL/well: 6.5 sec., 384 wells, 10 μL cass, 10 μL/well: 8 sec., 384 wells, 1 μL cass, 1 μL/well: 6 sec., 1536 wells, 1 μL cass, 1 μL/well: 21 s |
Electrical Requirements | 90 W max. |
Depth (English) | 11.8 in. |
Width (English) | 17.2 in. |
For Use With (Application) | Primary/secondary screening assays, compound storage, genomics and proteomics research, ELISA, cell based washing, fixing, staining, volume or concentration normalization |
Width (Metric) | 43.5 cm |
Accuracy | ±5% at 1 μL, ±2.0% at 5 μL, ±2.0% at 10 μL |
Precision | <5% CV at 1 μL, <2.5% CV at 5 μL, <2.0% CV at 10 μL |
For Use With (Equipment) | BioStack and third party automation compatible, BioSpa 8 Automated Incubator compatible |
Product Type | Microplate Dispenser |
Depth (Metric) | 29.2 cm |
Voltage | 100/240 V |
Weight (English) | 14.3 lb. |
Software | LHC2 Software, LHC2 Secure for 21 CFR Part 11 compliance (option), SiLA Compliant driver (option) |
Certifications/Compliance | CE and TUV marked, RoHS compliant |
Interface | Color Touchscreen |
Weight (Metric) | 6.5 kg |
Height (English) | 8 in. |
Plate Type | Dispensing (peri pump and syringe): 96-, 384- and 1536-well standard, deep and PCR plates; 6-. 12-, 24- and 48-well plates (dispense tip configurable); Washing: 96-. 384-well standard plates; 6-, 12-, 24-,and 48-well plates (with compatible manifold configuration) |
Agilent BioTek MultiFlo FX Multi-Mode Dispenser, IVD PROMO
MultiFlo FX is an automated multi-mode reagent dispenser for 6- to 1536-well microplates. For In Vitro Diagnostic Use.
Height (Metric) | 20.3 cm |
---|---|
Speed | 96 wells, 5 μL cass, 10 μL/well: 3 sec., 96 wells, 5 μL cass, 20 μL/well: 3.5 sec., 384 wells, 5 μL cass, 5 μL/well: 6.5 sec., 384 wells, 10 μL cass, 10 μL/well: 8 sec., 384 wells, 1 μL cass, 1 μL/well: 6 sec., 1536 wells, 1 μL cass, 1 μL/well: 21 s |
Electrical Requirements | 90 W max. |
Depth (English) | 11.8 in. |
Width (English) | 17.2 in. |
For Use With (Application) | Primary/secondary screening assays, compound storage, genomics and proteomics research, ELISA, cell based washing, fixing, staining, volume or concentration normalization |
Width (Metric) | 43.5 cm |
Accuracy | ±5% at 1 μL, ±2.0% at 5 μL, ±2.0% at 10 μL |
Precision | <5% CV at 1 μL, <2.5% CV at 5 μL, <2.0% CV at 10 μL |
For Use With (Equipment) | BioStack and third party automation compatible, BioSpa 8 Automated Incubator compatible |
Product Type | Microplate Dispenser |
Depth (Metric) | 29.2 cm |
Voltage | 100/240 V |
Weight (English) | 14.3 lb. |
Software | LHC2 Software, LHC2 Secure for 21 CFR Part 11 compliance (option), SiLA Compliant driver (option) |
Certifications/Compliance | CE and TUV marked, RoHS compliant |
Interface | Color Touchscreen |
Weight (Metric) | 6.5 kg |
Height (English) | 8 in. |
Plate Type | Dispensing (peri pump and syringe): 96-, 384- and 1536-well standard, deep and PCR plates; 6-. 12-, 24- and 48-well plates (dispense tip configurable); Washing: 96-. 384-well standard plates; 6-, 12-, 24-,and 48-well plates (with compatible manifold configuration) |
Agilent BioTek ELx405 Select Deep Well Microplate Washer
Greener Choice
Offers one or more environmental benefits itemized in the U.S. FTC Green Guides.
Learn More
Offers one or more environmental benefits itemized in the U.S. FTC Green Guides.
Learn More
The BioTek ELx405 Select is the first deep well plate washer that aspirates and dispenses into 96- and 384-well deep well and standard height microplates.
Height (Metric) | 25.4 cm |
---|---|
Depth (English) | 17 in. |
Fluid Delivery Type | One positive displacement pump |
Width (English) | 14 in. |
Temperature (English) Operating | 59°F to 86°F |
Width (Metric) | 35.6 cm |
Type | Microplate Washer |
Precision | ≤3% CV |
Depth (Metric) | 43.2 cm |
Wash Cycles | 1 to 10 |
Voltage | 100/240 V |
Humidity | 10 to 80%, noncondensing |
Soak Time | Programmable in minutes and seconds, up to 60 min. |
Amperage | <8 A |
Frequency | 50/60 Hz |
Residual Volume | ≤2 μL/well |
Weight (English) | 30 lb. |
Certifications/Compliance | CE and TUV marked. RoHS compliant. |
Temperature (Metric) Operating | 15°C to 30°C |
Height (English) | 10 in. |
Display | Backlit LCD |
Weight (Metric) | 13.5 kg |
Plate Type | 96-well Plate, 384-well Plate |
Agilent BioTek Cytation 5 Cell Imaging Multi-Mode Reader
Greener Choice
Offers one or more environmental benefits itemized in the U.S. FTC Green Guides.
Learn More
Offers one or more environmental benefits itemized in the U.S. FTC Green Guides.
Learn More
Cytation 5 combines automated microscopy and conventional microplate detection in a configurable, upgradable platform. For Research Use Only. Not for use in diagnostic procedures.
Height (Metric) | 44.5 cm |
---|---|
Depth (English) | 20.2 in. |
Width (English) | 16.4 in. |
Contrast Methods | Fluorescence, phase contrast, brightfield, high contrast brightfield, color brightfield and phase contrast |
Shaking | Linear, Orbital, Double Orbital |
Wattage | 250 W max. |
Objectives | 1.25x, 2.5x (2.25x eff), 2.5x (2.75x eff), 4x, 10x, 20x, 40x, 60x |
Absorbance Range | 230 to 999 nm, 1 nm increments |
Light Source | Xenon Flash; 100 mW 680 nm laser (Alpha Detection) |
Width (Metric) | 41.6 cm |
Dynamic Range | Absorbance: 0 to 4 OD; Fluorescence Intensity: 7 decades; Luminesence: >6 decades |
Reading Options | End point, kinetic, spectral scanning and well area scanning |
Accuracy | Monochromator: ±2 nm; Dispenser: ±1 μL or 2% |
Type | Cell Imaging Multi-Mode Microplate Reader |
No. of Dispensers | 2 Syringe Pumps |
Dispense Volume | 5 to 1000 μL, 1 μL increments |
For Use With (Equipment) | BioStack and Third Party Automation Compatible; BioSpa 8 Automated Incubator Compatible |
Depth (Metric) | 51.4 cm |
Weight (English) | 80 lb. |
Certifications/Compliance | CE and TUV marked. RoHS compliant. |
Camera | Sony CMOS, 16-bit Gray Scale, Standard or WFOV |
Temperature Range (Metric) | 4-Zone incubation to 65°C with Condensation Control (±0.2°C at 37°C) |
CO2 Concentration Range | 0 to 20% |
Weight (Metric) | 36.3 kg |
Height (English) | 17.5 in. |
Agilent BioTek Cytation 7 Cell Imaging Multi-Mode Reader
Greener Choice
Offers one or more environmental benefits itemized in the U.S. FTC Green Guides.
Learn More
Offers one or more environmental benefits itemized in the U.S. FTC Green Guides.
Learn More
BioTek Cytation 7 cell imaging multi-mode reader combines automated digital upright and inverted widefield microscopy with conventional multi-mode microplate reading. For Research Use Only. Not for use in diagnostic procedures.
Scan Speed | 96-wells: 11 s; 384-wells: 22 s |
---|---|
Shaking | Linear, Orbital, Double Orbital |
Light Source | Xenon Flash |
Wavelength | Luminescence 300 to 700 nm; Absorbance 230 to 999 nm, 1 nm increment |
Detection Method | Monochromator- based fluorescence intensity, luminescence and absorbance |
Reading Options | End point, kinetic, spectral scanning and well area scanning |
Sensitivity | Fluorescein 2.5 pM (0.25 fmol/well, 384-well plate) – top Fluorescein 4 pM (0.4 fmol/well, 384-well plate) – bottom; luminescence 20 amol ATP (flash) |
Type | Automated Microscope/Multi-Mode Microplate Reader |
No. of Dispensers | 2 Syringe Pumps |
Dispense Volume | 5 to 1000 μL, 1 μL increments |
Linearity | <1% from 0 to 3 OD |
Voltage | 100/240 V |
Bandwidth | 4 nm (230 to 285 nm), 8 nm (>285 nm) |
Frequency | 50/60 Hz |
Certifications/Compliance | CE and TUV marked. RoHS compliant. |
Wavelength Selection | Quad monochromators (top/bottom) |
Temperature Range (Metric) | 4-Zone incubation to 45°C with Condensation Control (±0.5°C at 37°C) |
Resolution | 0.0001 OD |
Plate Type | Monochromator: 6- to 384-well plates; Imaging: 6- to 1536-well plates |
Agilent BioTek 405 TS Touch Microplate Washer, IVD
Greener Choice
Offers one or more environmental benefits itemized in the U.S. FTC Green Guides.
Learn More
Offers one or more environmental benefits itemized in the U.S. FTC Green Guides.
Learn More
The BioTek 405 TS washer has an excellent reputation in life science research for outstanding results in many applications. For In Vitro Diagnostic Use.
Connections | USB (2) |
---|---|
Width (English) | 14 in. |
For Use With (Application) | ELISA, MSD assays, HCS immune cytochemistry, FLIPR Ca2+ flux, Cell-based assays, Magnetic and polystyrene bead processing (gene expression assays, cytokine assays), ELISPOT assays |
Width (Metric) | 35.6 cm |
Assays | ELISA, Cell-based assays (model dependent), Magnetic and polystyrene bead, Multiplex assays, Bead-based ELISAs, Filtration-to-waste processes |
Flow Rate | High to Low Flow; Optimized rates for cell assays |
Bottles | Optional (4 or 10 L) |
Wash Cycles | 1 to 250 |
Voltage | 100/240 V |
Certifications/Compliance | CE and TUV marked. RoHS compliant. |
Height (English) | 10 in. |
Weight (Metric) | With internal buffer switching - 16.5 kg; Without - 13.5 kg |
Wash Volume | 25 to 3000 μL/well |
Height (Metric) | 25.4 cm |
Depth (English) | 17 in. |
Fluid Delivery Type | Internal positive displacement pump |
Automation Interface | High Resolution LED Backlit Touch Screen |
Type | Microplate Washer |
Washing Speed | 96-wells: 300 μL/well, 3 cycles: <30 s; 384-wells: 100 μL/well, 3 cycles: <80 s; 384-wells: 400 μL/well, 1 cycle: <20 s |
Dispense Volume | 25 to 3000 μL/well (192-pin manifold); 50 to 3000 μL/well (96-pin manifold) |
Precision | <3% CV: 300 μL/well (96-well washing); <4% CV: 80 μL/well (384-well washing) |
Depth (Metric) | 43.2 cm |
Soak Time | Programmable in minutes and seconds, up to 60 min. |
Frequency | 50/60 Hz |
Residual Volume | ≤2 μL/well |
Weight (English) | With internal buffer switching - 36 lb.; Without - 30 lb. |
Display | LED |
Plate Type | 96- and 384-well, Low Profile and Standard, Solid and Filter Bottom (Option) Plates |
Agilent BioTek 405LS Microplate Washer, IVD
Greener Choice
Offers one or more environmental benefits itemized in the U.S. FTC Green Guides.
Learn More
Offers one or more environmental benefits itemized in the U.S. FTC Green Guides.
Learn More
The BioTek 405 LS washer has a well earned reputation as an economical plate washer that offers excellent performance and reliability. For In Vitro Diagnostic Use.
Height (Metric) | 25.4 cm |
---|---|
Depth (English) | 17 in. |
Width (English) | 14 in. |
Width (Metric) | 35.6 cm |
Flow Rate | High to Low Flow; Optimized rates for cell assays |
Type | Microplate Washer |
Bottles | Optional (4 or 10 L) |
Washing Speed | 96-wells: 300 μL/well, 3 cycles: <30 s; 384-wells: 100 μL/well, 3 cycles: <80 s; 384-wells: 400 μL/well, 1 cycle: <20 s |
Dispense Volume | 25 to 3000 μL/well |
Precision | <3% CV: 300 μL/well (96-well washing); <4% CV: 80 μL/well (384-well washing) |
Depth (Metric) | 43.2 cm |
Wash Cycles | 1 to 250 |
Voltage | 100/240 V |
Humidity | 10 to 85%, noncondensing |
Soak Time | Programmable in minutes and seconds, up to 60 min. |
Increments | 1 μL |
Frequency | 50/60 Hz |
Weight (English) | With internal buffer switching - 36 lb.; Without - 30 lb. |
Certifications/Compliance | CE and TUV marked. RoHS compliant. |
Height (English) | 10 in. |
Weight (Metric) | With internal buffer switching - 16.5 kg; Without - 13.5 kg |
Plate Type | 96- and 384-well, Low Profile and Standard, Solid and Filter Bottom (Pore Sizes 0.45 to 1.2 μm) Plates |
Agilent BioTek ELx405 Select Microplate Washer, IVD
Greener Choice
Offers one or more environmental benefits itemized in the U.S. FTC Green Guides.
Learn More
Offers one or more environmental benefits itemized in the U.S. FTC Green Guides.
Learn More
The BioTek ELx405 Select is the first deep well plate washer that aspirates and dispenses into 96- and 384-well deep well and standard height microplates. For In Vitro Diagnostic Use.
Height (Metric) | 25.4 cm |
---|---|
Depth (English) | 17 in. |
Fluid Delivery Type | One positive displacement pump |
Width (English) | 14 in. |
Temperature (English) Operating | 59°F to 86°F |
Width (Metric) | 35.6 cm |
Type | Microplate Washer |
Precision | ≤3% CV |
Depth (Metric) | 43.2 cm |
Wash Cycles | 1 to 10 |
Voltage | 100/240 V |
Humidity | 10 to 80%, noncondensing |
Soak Time | Programmable in minutes and seconds, up to 60 min. |
Amperage | <8 A |
Frequency | 50/60 Hz |
Residual Volume | ≤2 μL/well |
Weight (English) | 30 lb. |
Certifications/Compliance | CE and TUV marked. RoHS compliant. |
Temperature (Metric) Operating | 15°C to 30°C |
Height (English) | 10 in. |
Display | Backlit LCD |
Weight (Metric) | 13.5 kg |
Agilent BioTek Synergy Neo2 Hybrid Multi-Mode Microplate Reader, IVD
Greener Choice
Offers one or more environmental benefits itemized in the U.S. FTC Green Guides.
Learn More
Offers one or more environmental benefits itemized in the U.S. FTC Green Guides.
Learn More
BioTek Synergy Neo2 multi-mode microplate reader features patented Hybrid Technology, with independent optical paths that ensure excellent performance in all detection modes. For In Vitro Diagnostic Use.
Height (Metric) | 41 cm |
---|---|
Depth (English) | 20.7 in. |
Width (English) | 15.4 in. |
Shaking | Linear, Orbital, Double Orbital |
Software Type | Gen5 Data Analysis Software,Gen5 Secure for 21 CFR Part 11 compliance (option) |
Absorbance Range | 230 to 999 nm, 1 nm increments |
Incubator Temperature | ±0.2°C at 37°C |
Light Source | Xenon Flash |
For Use With (Application) | HTS, biomarker, nucleic acid, protein quantification, rapid kinetics, HTRF, SNP, TR FRET, BRET, Cytokines, ELISA, FP |
Width (Metric) | 39 cm |
Dynamic Range | 0 to 4 OD |
Detector Type | Photodiode,Fluorescence/Alpha: Dual PMT or Single PMT (option) |
Accuracy | Monochromator: ±2 nm; <1% at 2.0 OD, <3% at 3.0 OD |
Dispense Volume | 5 to 1000 μL, 1 μL increments |
Product Type | Multi-Mode Microplate Reader |
Linearity | ≤1% from 0 to 3 OD |
Depth (Metric) | 52.5 cm |
Weight (English) | 78 lb. |
Certifications/Compliance | CE and TUV marked. RoHS compliant. |
Power Consumption | 250 W max. |
CO2 Concentration Range | 0 to 20% |
Weight (Metric) | 35 kg |
Resolution | 0.0001 OD |
Height (English) | 16.1 in. |