UserName
Viewing profile as user:
MP Biomedicals FastPrep Sample Preparation Products
MP Biomedicals FastPrep Sample Preparation Products
Explore the complete collection of MP Biomedicals™ FastPrep™ homogenizers and accessories, lysing matrices, and nucleic acid extraction kits to help optimize your workflow.
FastPrep homogenizers offer rapid and effective bead-beating action that can lyse and homogenize most plant and animal tissue and other organic samples. Then extract, purify, and further analyze the released nucleic acids with FastPrep DNA and RNA Isolation kits.
To help you choose the right homogenizing products to fit your needs:
  • Determine the number of samples being processed
    Based on the sample sizes and volumes, choose a homogenizer and identify the adapters or other accessories needed for your samples
  • Choose an appropriate lysing matrix and extraction kit
    Make a selection based on the type of sample (soft tissue, woody stems, microbes, etc.) and sample volume
  • Review the available FastPrep Starter Kits
    One of the starter kits may be suitable for your application
Find FastPrep Homogenizers and Matrices
Select a FastPrep Homogenizer
The bead-beating technology of FastPrep systems completely lyses virtually any sample in seconds. Choose a legacy benchtop instrument, a version that features increased power levels, a high-throughput homogenizer, or a hand-held model for field or remote work.
Shop Homogenizer Products
Identify Sample Holder Adapters
FastPrep Sample Holder Adapters help accommodate a range of sample quantities, types, and volumes to meet your sample preparation needs. From 2 mL tubes to 96-deep well plates, our broad selection of interchangeable adapters adds flexibility to your workflow.
Shop Homogenizer Adapters
FastPrep Sample Holder Adapters
Select a Lysing Matrix
Lysing matrix tubes contain beads of different sizes, shapes, and materials to homogenize various sample types. Available in a range of sizes to accommodate most samples, choose a 96-well format or 2 mL, 4.5 mL, 15 mL, or 50 mL tubes to prepare cells, tissues, environmental samples, insects, plants, and more.
Shop Lysing Matrices
Choose DNA and RNA Isolation Kits
After your samples have been lysed and homogenized, use FastRNA and FastDNA kits to extract and purify the nucleic acids for downstream applications. These kits have been developed to produce high yields of DNA and RNA.
Shop Isolation Kits
DNA and RNA Isolation Kits
Select a FastPrep Starter Package
FastPrep Starter Packages are designed for labs that deal with more than one sample type. Each Starter Package includes:
  • FastPrep-24 5G homogenizer
  • Assorted lysing matrix tubes
  • Adapter(s)
  • Nucleic acid extraction and purification reagents and kits
Shop Starter Packages
FastPrep Starter Package