UserName
Viewing profile as user:
Thermo Scientific Dry Baths/Block Heaters
Thermo Scientific Dry Baths Block Heaters BNR
Digital Dry Bath Block Heater
Digital Dry Bath/Block Heater
Increase lab versatility with Thermo Scientific™ Digital Dry Baths/Block Heaters, which offer a range of configurations with interchangeable modular blocks for a variety application needs.
Features Include:
  • Digital controls and display of time and temperature
  • Precise temperature control with PID circuit
  • Temperature calibration enables to offset the temperature to desired value
  • Timer allows the user to accurately monitor the heating time
Applications include:
  • Sample preparation
  • Enzyme preservation
  • Enzyme-substrate reactions
  • DNA amplification
  • Electrophoresis gel degeneration
  • Serum coagulation
Thermo Scientific Dry Baths/Block Heaters Brochure
Reliability you can depend on. Thermo Scientific™ versatile dry baths / block heaters offer a wide range of blocks to accommodate various tubes.
Touch Screen Bath Block Heater
Touch Screen Dry Bath Block Heater
Touch Screen Dry Bath/Block Heater
Thermo Scientific™ Touch Screen Dry Bath /Block Heater is designed to achieve precise temperature stability and uniformity with an easy-to-use touch screen.
Features Include:
  • Temperature deviation adjustment
  • Precise temperature control with PID circuit
  • Temperature calibration enables to offset the temperature to desired value
  • Timer enables the user to accurately monitor the heating time
  • Optional temperature for better temperature accuracy
  • Programmable
Applications include:
  • Sample preparation
  • Enzyme preservation
  • Enzyme-substrate reactions
  • DNA amplification
  • Electrophoresis gel degeneration
  • Serum coagulation
Thermo Scientific Dry Baths/Block Heaters Brochure
Reliability you can depend on. Thermo Scientific™ versatile dry baths / block heaters offer a wide range of blocks to accommodate various tubes.
Touch Screen Bath Block Heater
Blocks
Blocks