UserName
Viewing profile as user:
Safe-Cote Bottles
Safe-Cote Bottles
Fisher Chemical Products in Safe-Cote Bottles
The following Fisher Chemical products are packaged in Safe-Cote™ PVC bottles that provide the purity of glass and most of the benefits of plastic for storing and dispensing chemicals. If they break, glass fragments and liquids are more likely to remain trapped.
Fisher Chemical products also include the following packaging features:
  • Innovative safety bottle with FisherLOCK™ tamper-evident cap
  • Convenient storing and dispensing
  • Packaged in the 100% recyclable Styrofoam™-free EcoSafPak™
Safe-Cote Demonstration
See a demonstration of a Safe-Cote bottle vs. a regular bottle to see how Safe-Cote can help protect your chemicals, your lab and most importantly, your people.
SolventGradeSizeCat No.
1-Butanol
HPLC
1L – 4L
A383SK
2-Propanol
OPTIMA
4L
A464SK-4
2-Propanol
HPLC
1L – 4L
A451SK
2-Propanol
Certified ACS Plus
4L
A416SK-4
Acetone
HPLC
1L – 4L
A949SK
Acetone
Certified ACS
4L
A18SK-4
Acetonitrile
OPTIMA
4L
A996SK4
Acetonitrile
HPLC
1L – 4L
A998SK
Chloroform (Approx. 0.75% Ethanol as Preservative)
Certified ACS
4L
C298SK-4
Chloroform (Approx. 50ppm Pentene as Preservative)
HPLC
1L – 4L
C607SK
Chloroform (Approx.0.75% Ethanol as Preservative)
HPLC
1L – 4L
C606SK
Chloroform (Approx.0.75% Ethanol as Preservative)
HPLC
4L
C606SK-4
Cyclohexane
HPLC
1L – 4L
C620SK
Ethyl Acetate
OPTIMA
4L
E196SK-4
Ethyl Acetate
HPLC
1L – 4L
E195SK
Ethyl Acetate
Certified ACS
4L
E145SK-4
Ethyl Alcohol Denatured
Proprietary Solvent
4L
A407SK-4
Heptane
HPLC
1L – 4L
H350SK
Hexanes
OPTIMA
4L
H303SK-4
Hexanes
HPLC
1L – 4L
H302SK
Hexanes
Certified ACS
4L
H292SK-4
Isooctane
HPLC
1L – 4L
O296SK
Methanol
OPTIMA
4L
A454SK-4
Methanol
HPLC
1L – 4L
A452SK
Methanol
Certified ACS
4L
A412SK-4
Methanol
Spectranalyzed
4L
A408SK-4
Methylene Chloride
OPTIMA
4L
D151SK-4
Methylene Chloride (Not Stabilized)
HPLC
1L – 4L
D150SK
Methylene Chloride (Stabilized)
HPLC
1L – 4L
D143SK
Methylene Chloride (Stabilized)
Certified ACS
4L
D37SK-4
Methylene Chloride (With Cyclohexene Preservative)
HPLC
4L
D138SK-4
n-Butyl Chloride
HPLC
4L
B429SK-4
n-Hexane, 95%
OPTIMA
4L
H306SK-4
Pentane
HPLC
1L – 4L
P399SK
Petroleum Ether
OPTIMA
4L
E120SK-4
Petroleum Ether
Certified ACS
4L
E139SK-4
Tetrahydrofuran
OPTIMA
4L
T427SK-4
Tetrahydrofuran
HPLC
1L – 4L
T425SK-1
Tetrahydrofuran
Certified
4L
T397SK-4
Toluene
OPTIMA
4L
T291SK-4
Toluene
HPLC
1L – 4L
T290SK
Toluene
Certified ACS
4L
T324SK-4
Toluene
Scintanalyzed
4L
T313SK-4
Water
OPTIMA
4L
W7SK-4
Water
HPLC
1L – 4L
W5SK
Xylenes
Certified ACS
4L
X5SK-4