Assay Details
Target Gene Details
Entrez Gene ID: | 1050 |
Gene Name: | CCAAT/enhancer binding protein alpha |
Gene Aliases: |
C/EBP-alpha, CEBP |
Location: |
Chr.19:33299934-33302564 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CEBPA | NM_001285829.1 | 1 | 1509 | NP_001272758.1 |
| NM_001287424.1 | 1 | 1509 | NP_001274353.1 | |
| NM_001287435.1 | 1 | 1509 | NP_001274364.1 | |
| NM_004364.4 | 1 | 1509 | NP_004355.2 | |
| BC063874.1 | 2 | 1038 | AAH63874.1 | |
| X87248.1 | 1 | 1422 | CAA60698.1 | |
| Y11525.1 | 1 | 1509 | CAA72289.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833808 | Chr.19:33130887 - 33350702 on Build GRCh38 | Loss |
|
| nsv2463 | Chr.19:33294939 - 33313582 on Build GRCh38 | Deletion |
|
| esv2758755 | Chr.19:33286168 - 34090656 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map