Assay Details
Target Gene Details
Entrez Gene ID: | 65125 |
Gene Name: | WNK lysine deficient protein kinase 1 |
Gene Aliases: |
HSAN2, HSN2, KDP, PPP1R167, PRKWNK1, PSK, p65 |
Location: |
Chr.12:752552-911452 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| WNK1 | NM_001184985.1 | 1 | 675 | NP_001171914.1 |
| NM_014823.2 | 1 | 675 | NP_055638.2 | |
| NM_018979.3 | 1 | 675 | NP_061852.3 | |
| NM_213655.4 | 1 | 675 | NP_998820.3 | |
| XM_006719003.2 | 1 | 1000 | XP_006719066.1 | |
| XM_011520997.2 | 1 | 1046 | XP_011519299.1 | |
| XM_011520998.2 | 1 | 1000 | XP_011519300.1 | |
| XM_011520999.2 | 1 | 1000 | XP_011519301.1 | |
| XM_011521000.2 | 1 | 1000 | XP_011519302.1 | |
| XM_011521001.2 | 1 | 1000 | XP_011519303.1 | |
| XM_011521002.2 | 1 | 1000 | XP_011519304.1 | |
| XM_011521003.2 | 1 | 1000 | XP_011519305.1 | |
| XM_011521004.2 | 1 | 1000 | XP_011519306.1 | |
| XM_011521006.2 | 1 | 1000 | XP_011519308.1 | |
| XM_011521007.2 | 1 | 1000 | XP_011519309.1 | |
| XM_011521008.2 | 1 | 1000 | XP_011519310.1 | |
| XM_011521009.2 | 1 | 1000 | XP_011519311.1 | |
| XM_017019834.1 | 1 | 1000 | XP_016875323.1 | |
| XM_017019835.1 | 1 | 1000 | XP_016875324.1 | |
| XM_017019836.1 | 1 | 1000 | XP_016875325.1 | |
| XM_017019837.1 | 1 | 1000 | XP_016875326.1 | |
| XM_017019838.1 | 1 | 1000 | XP_016875327.1 | |
| XM_017019839.1 | 1 | 1000 | XP_016875328.1 | |
| AF061944.1 | 1 | 675 | AAF31483.1 | |
| AJ296290.1 | CAC15059.1 | |||
| BC141881.1 | ||||
| JQ358908.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv2251n54 | Chr.12:553282 - 1101718 on Build GRCh38 | Gain |
|
| esv2760688 | Chr.12:746699 - 767080 on Build GRCh38 | Loss |
|
| dgv2253n54 | Chr.12:741122 - 859323 on Build GRCh38 | Loss |
|
| nsv556887 | Chr.12:523074 - 758338 on Build GRCh38 | Gain |
|
| dgv1335n100 | Chr.12:739562 - 764851 on Build GRCh38 | Loss |
|
| nsv468959 | Chr.12:741447 - 813242 on Build GRCh38 | Loss |
|
| esv2759875 | Chr.12:225844 - 881378 on Build GRCh38 | Gain+Loss |
|
| nsv556893 | Chr.12:628064 - 873155 on Build GRCh38 | Gain |
|
| nsv518946 | Chr.12:745165 - 852170 on Build GRCh38 | Loss |
|
| nsv1119925 | Chr.12:739873 - 764441 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map