Assay Details
Target Gene Details
Entrez Gene ID: | 6557 |
Gene Name: | solute carrier family 12 member 1 |
Gene Aliases: |
BSC1, NKCC2 |
Location: |
Chr.15:48206301-48304078 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC12A1 | NM_000338.2 | 2 | 540 | NP_000329.2 |
| NM_001184832.1 | 2 | 540 | NP_001171761.1 | |
| XM_005254606.1 | 2 | 540 | XP_005254663.1 | |
| AK295375.1 | ||||
| AK298312.1 | 2 | 539 | ||
| BC040138.2 | 1 | 510 | AAH40138.1 | |
| DA632985.1 | ||||
| EF559316.2 | 1 | 338 | ||
| U58130.1 | 1 | 343 | AAB07364.1 |
More Information
Set Membership: |
|

Genomic Map