Assay Details
Target Gene Details
Entrez Gene ID: | 1417 |
Gene Name: | crystallin beta B3 |
Gene Aliases: |
CATCN2, CRYB3, CTRCT22 |
Location: |
Chr.22:25199312-25207617 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CRYBB3 | NM_004076.4 | 4 | 360 | NP_004067.1 |
| XM_011529902.2 | 4 | 898 | XP_011528204.1 | |
| XM_017028599.1 | 4 | 898 | XP_016884088.1 | |
| BC069479.1 | 4 | 330 | AAH69479.2 | |
| BC102021.1 | 4 | 317 | AAI02022.1 | |
| BC102022.1 | 4 | 317 | AAI02023.1 | |
| BC107482.1 | 4 | 317 | AAI07483.1 | |
| BM711382.1 | 4 | 360 | ||
| CR456427.1 | 3 | 311 | CAG30313.1 | |
| U71216.1 | AAC50972.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv469659 | Chr.22:25202943 - 25366882 on Build GRCh38 | Gain+Loss |
|
| nsv834162 | Chr.22:25149393 - 25313043 on Build GRCh38 | Loss |
|
| dgv24e196 | Chr.22:25116284 - 25403903 on Build GRCh38 | Duplication |
|
| esv2758829 | Chr.22:24970834 - 25650546 on Build GRCh38 | Gain+Loss |
|
| nsv428388 | Chr.22:25153850 - 25435228 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map