Assay Details
Target Gene Details
Entrez Gene ID: | 7045 |
Gene Name: | transforming growth factor beta induced |
Gene Aliases: |
BIGH3, CDB1, CDG2, CDGG1, CSD, CSD1, CSD2, CSD3, EBMD, LCD1 |
Location: |
Chr.5:136028895-136063818 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TGFBI | NM_000358.2 | NP_000349.1 | ||
| AK093916.1 | ||||
| AK094055.1 | ||||
| AK094581.1 | ||||
| AK222833.1 | BAD96553.1 | |||
| AK298603.1 | ||||
| AK303643.1 | ||||
| BC000097.2 | AAH00097.1 | |||
| BC004972.1 | AAH04972.1 | |||
| BT009820.1 | AAP88822.1 | |||
| DA051240.1 | ||||
| KU178357.1 | ||||
| KU178358.1 | ||||
| M77349.1 | AAA61163.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2759380 | Chr.5:135829912 - 136139310 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map