Assay Details
Target Gene Details
Entrez Gene ID: | 6505 |
Gene Name: | solute carrier family 1 member 1 |
Gene Aliases: |
DCBXA, EAAC1, EAAT3, SCZD18 |
Location: |
Chr.9:4490427-4587469 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC1A1 | NM_004170.5 | 1 | 278 | NP_004161.4 |
| XM_011518007.1 | 1 | 229 | XP_011516309.1 | |
| XM_011518010.1 | 1 | 229 | XP_011516312.1 | |
| XM_017015042.1 | 1 | 229 | XP_016870531.1 | |
| XM_017015043.1 | 1 | 239 | XP_016870532.1 | |
| AB008536.1 | 1 | 216 | BAB83767.1 | |
| AK313013.1 | 1 | 179 | ||
| BC033040.1 | 1 | 264 | AAH33040.1 | |
| DA643262.1 | 1 | 278 | ||
| U03506.1 | AAA50430.1 | |||
| U06469.1 | 1 | 64 | AAA53215.1 | |
| U08989.1 | 1 | 188 | AAA68628.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1016929 | Chr.9:4219016 - 4545685 on Build GRCh38 | Gain |
|
| nsv1022870 | Chr.9:4416046 - 4611901 on Build GRCh38 | Loss |
|
| nsv524574 | Chr.9:4219259 - 4543307 on Build GRCh38 | Gain |
|
| nsv1022493 | Chr.9:4490134 - 4604192 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map