Assay Details
Target Gene Details
Entrez Gene ID: | 208 |
Gene Name: | AKT serine/threonine kinase 2 |
Gene Aliases: |
HIHGHH, PKBB, PKBBETA, PRKBB, RAC-BETA |
Location: |
Chr.19:40230317-40285531 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 16 - Exon 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| AKT2 | NM_001243027.2 | NP_001229956.1 | ||
| NM_001243028.2 | NP_001229957.1 | |||
| NM_001626.5 | NP_001617.1 | |||
| XM_011526614.1 | XP_011524916.1 | |||
| XM_011526615.1 | XP_011524917.1 | |||
| XM_011526616.1 | XP_011524918.1 | |||
| XM_011526618.1 | XP_011524920.1 | |||
| XM_011526619.1 | XP_011524921.1 | |||
| XM_011526620.1 | XP_011524922.1 | |||
| XM_011526622.2 | XP_011524924.1 | |||
| XM_017026470.1 | XP_016881959.1 | |||
| XM_017026471.1 | XP_016881960.1 | |||
| AK054771.1 | 1 | 1264 | ||
| AK055779.1 | ||||
| AK122839.1 | 2 | 2314 | ||
| AK294453.1 | ||||
| AK308006.1 | ||||
| AK308906.1 | ||||
| AK314619.1 | ||||
| BC022779.1 | AAH22779.1 | |||
| BC032709.1 | ||||
| BC063421.1 | AAH63421.1 | |||
| BC120994.1 | ||||
| BC120995.1 | ||||
| KU159425.1 | ||||
| M77198.1 | AAA36585.1 | |||
| M95936.1 | AAA58364.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1062755 | Chr.19:40231556 - 40252328 on Build GRCh38 | Loss |
|
| esv3644345 | Chr.19:40203124 - 40288712 on Build GRCh38 | Gain |
|
| esv2758761 | Chr.19:40106513 - 40333613 on Build GRCh38 | Gain |
|
| esv3644344 | Chr.19:40191059 - 40328585 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map