Assay Details
Target Gene Details
Entrez Gene ID: | 5313 |
Gene Name: | pyruvate kinase, liver and RBC |
Gene Aliases: |
PK1, PKL, PKR, PKRL, RPK |
Location: |
Chr.1:155289293-155301434 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PKLR | NM_000298.5 | NP_000289.1 | ||
| NM_181871.3 | NP_870986.1 | |||
| XM_006711386.3 | XP_006711449.1 | |||
| XM_011509640.2 | XP_011507942.1 | |||
| XM_017001493.1 | XP_016856982.1 | |||
| AB015983.1 | BAA31706.1 | |||
| AK225947.1 | ||||
| AK298399.1 | ||||
| BC025737.1 | AAH25737.1 | |||
| M15465.1 | AAA60104.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv3032 | Chr.1:155285619 - 155318185 on Build GRCh38 | Insertion |
|
| nsv831636 | Chr.1:155249432 - 155475073 on Build GRCh38 | Loss |
|
| esv33869 | Chr.1:155253492 - 155948170 on Build GRCh38 | Loss |
|
| nsv952214 | Chr.1:155279610 - 155304509 on Build GRCh38 | Deletion |
|
| dgv3n68 | Chr.1:155122502 - 155343618 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map