Assay Details
Target Gene Details
Entrez Gene ID: | 26227 |
Gene Name: | phosphoglycerate dehydrogenase |
Gene Aliases: |
3-PGDH, 3PGDH, HEL-S-113, NLS, NLS1, PDG, PGAD, PGD, PGDH, PHGDHD, SERA |
Location: |
Chr.1:119711737-119744226 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 9 - Intron 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PHGDH | NM_006623.3 | NP_006614.2 | ||
| XM_011541226.1 | XP_011539528.1 | |||
| XM_011541227.2 | XP_011539529.1 | |||
| XM_011541228.1 | XP_011539530.1 | |||
| XM_011541231.2 | XP_011539533.1 | |||
| AF171236.1 | AAD51414.1 | |||
| AF171237.1 | AAD51415.1 | |||
| AK093306.1 | ||||
| AK309662.1 | ||||
| AK310132.1 | ||||
| AK315360.1 | ||||
| BC000303.2 | AAH00303.1 | |||
| BC001349.2 | AAH01349.1 | |||
| BC011262.1 | AAH11262.1 | |||
| CR456795.1 | CAG33076.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv831215 | Chr.1:119723826 - 119905846 on Build GRCh38 | Gain |
|
| nsv524681 | Chr.1:118890895 - 121570001 on Build GRCh38 | Gain |
|
| nsv428201 | Chr.1:119724690 - 120154589 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map