Assay Details
Target Gene Details
Entrez Gene ID: | 3831 |
Gene Name: | kinesin light chain 1 |
Gene Aliases: |
KLC, KNS2, KNS2A |
Location: |
Chr.14:103629188-103701551 on Build GRCh38 |
Assay Gene Location: | Within Intron 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KLC1 | NM_001130107.1 | NP_001123579.1 | ||
| NM_182923.3 | NP_891553.2 | |||
| AB451299.1 | ||||
| AB451431.1 | ||||
| AY180165.3 | AAO62550.1 | |||
| AY180168.1 | AAO62553.1 | |||
| BM988840.1 | ||||
| L04733.1 | AAA16576.1 |
Target Gene Details
Entrez Gene ID: | 7517 |
Gene Name: | X-ray repair cross complementing 3 |
Gene Aliases: |
CMM6 |
Location: |
Chr.14:103697611-103715486 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| XRCC3 | NM_001100118.1 | 9 | 2352 | NP_001093588.1 |
| NM_001100119.1 | 10 | 2446 | NP_001093589.1 | |
| NM_005432.3 | 10 | 2409 | NP_005423.1 | |
| XM_005268046.2 | 9 | 2446 | XP_005268103.1 | |
| XM_011537138.2 | 10 | 2392 | XP_011535440.1 | |
| AF035586.1 | 9 | 2337 | AAC05368.1 | |
| AK022829.1 | 10 | 2338 | ||
| AK023646.1 | 10 | 2382 | ||
| AK124498.1 | 8 | 2826 | ||
| AK126706.1 | 5 | 3478 | ||
| BC001036.1 | 10 | 2381 | AAH01036.1 | |
| BC002949.1 | 1 | 1156 | AAH02949.1 | |
| BC011725.2 | 9 | 2312 | AAH11725.1 | |
| BX161398.1 | 9 | 2321 | CAD61884.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3635603 | Chr.14:103624778 - 103714856 on Build GRCh38 | Gain |
|
| nsv566023 | Chr.14:103595309 - 103731914 on Build GRCh38 | Loss |
|
| nsv952260 | Chr.14:103697564 - 103723263 on Build GRCh38 | Deletion |
|
| nsv470673 | Chr.14:103695926 - 103753612 on Build GRCh38 | Loss |
|
| nsv1054574 | Chr.14:103210111 - 105365114 on Build GRCh38 | Loss |
|
| esv2760013 | Chr.14:103681188 - 103946476 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map