Assay Details
Target Gene Details
Entrez Gene ID: | 2289 |
Gene Name: | FK506 binding protein 5 |
Gene Aliases: |
AIG6, FKBP51, FKBP54, P54, PPIase, Ptg-10 |
Location: |
Chr.6:35573585-35728583 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 12 - Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FKBP5 | NM_001145775.2 | NP_001139247.1 | ||
| NM_001145776.1 | NP_001139248.1 | |||
| NM_004117.3 | NP_004108.1 | |||
| AF194172.1 | AAL54872.1 | |||
| AK222807.1 | BAD96527.1 | |||
| AK302704.1 | ||||
| AK303356.1 | ||||
| AK312422.1 | ||||
| BC042605.1 | AAH42605.1 | |||
| BC111050.1 | AAI11051.1 | |||
| BX647390.1 | ||||
| U42031.1 | AAA86245.1 | |||
| U71321.1 | AAC51189.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3608684 | Chr.6:35554207 - 35601118 on Build GRCh38 | Loss |
|
| esv2759420 | Chr.6:35495964 - 35838727 on Build GRCh38 | Gain |
|
| esv3608680 | Chr.6:35521204 - 35600518 on Build GRCh38 | Gain |
|
| nsv602923 | Chr.6:35534425 - 35624967 on Build GRCh38 | Gain |
|
| dgv5957n100 | Chr.6:35534827 - 35611598 on Build GRCh38 | Gain |
|
| nsv602924 | Chr.6:35537534 - 35782522 on Build GRCh38 | Gain |
|
| dgv1176e214 | Chr.6:35552228 - 35601118 on Build GRCh38 | Gain |
|
| nsv7889 | Chr.6:35527726 - 35598611 on Build GRCh38 | Gain |
|
| esv2761067 | Chr.6:35503469 - 35605468 on Build GRCh38 | Gain |
|
| dgv5956n100 | Chr.6:35508785 - 35611598 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map