Assay Details
Target Gene Details
Entrez Gene ID: | 2070 |
Gene Name: | EYA transcriptional coactivator and phosphatase 4 |
Gene Aliases: |
CMD1J, DFNA10 |
Location: |
Chr.6:133240340-133532120 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| EYA4 | NM_001301012.1 | 1 | 191 | NP_001287941.1 |
| NM_001301013.1 | 1 | 191 | NP_001287942.1 | |
| NM_004100.4 | 1 | 333 | NP_004091.3 | |
| NM_172103.3 | 1 | 333 | NP_742101.2 | |
| NM_172105.3 | 1 | 333 | NP_742103.1 | |
| XM_005266851.4 | 1 | 40 | XP_005266908.1 | |
| XM_005266853.4 | 1 | 40 | XP_005266910.1 | |
| XM_017010368.1 | 2 | 560 | XP_016865857.1 | |
| XM_017010369.1 | 1 | 40 | XP_016865858.1 | |
| XM_017010370.1 | 1 | 40 | XP_016865859.1 | |
| XM_017010371.1 | 1 | 40 | XP_016865860.1 | |
| XM_017010372.1 | 1 | 40 | XP_016865861.1 | |
| XM_017010373.1 | 1 | 40 | XP_016865862.1 | |
| XM_017010374.1 | 1 | 339 | XP_016865863.1 | |
| XM_017010375.1 | 1 | 313 | XP_016865864.1 | |
| AK295798.1 | 1 | 340 | ||
| AK299378.1 | 1 | 105 | ||
| AK301950.1 | 2 | 431 | ||
| BC041063.1 | 1 | 272 | AAH41063.1 | |
| BX490250.1 | 1 | 333 | ||
| DA760903.1 | 1 | 105 | ||
| Y17114.1 | 1 | 315 | CAA76636.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv5483 | Chr.6:133233165 - 133278200 on Build GRCh38 | Deletion |
|
| nsv470862 | Chr.6:133171368 - 133279264 on Build GRCh38 | Loss |
|
| esv2759470 | Chr.6:133123580 - 133306226 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map