Assay Details
Target Gene Details
Entrez Gene ID: | 4982 |
Gene Name: | TNF receptor superfamily member 11b |
Gene Aliases: |
OCIF, OPG, PDB5, TR1 |
Location: |
Chr.8:118923557-118952144 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TNFRSF11B | NM_002546.3 | 2 | 680 | NP_002537.3 |
| AB002146.1 | 2 | 357 | BAA25910.1 | |
| AF134187.1 | 1 | 295 | AAF20168.1 | |
| AK223155.1 | 2 | 419 | BAD96875.1 | |
| AK308524.1 | 2 | 420 | ||
| AK313710.1 | 2 | 774 | ||
| BC030155.2 | 2 | 578 | AAH30155.1 | |
| BM748946.1 | ||||
| U94332.1 | 2 | 451 | AAB53709.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2759639 | Chr.8:118931070 - 119263510 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map