Assay Details
Target Gene Details
Entrez Gene ID: | 11141 |
Gene Name: | interleukin 1 receptor accessory protein like 1 |
Gene Aliases: |
IL1R8, IL1RAPL, MRX10, MRX21, MRX34, OPHN4, TIGIRR-2 |
Location: |
Chr.X:28587564-29956350 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| IL1RAPL1 | NM_014271.3 | NP_055086.1 | ||
| AF181284.1 | 1 | 113 | AAF59411.1 | |
| AJ243874.1 | CAB56046.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv6841 | Chr.X:28580318 - 28610714 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map