Assay Details
Target Gene Details
Entrez Gene ID: | 3662 |
Gene Name: | interferon regulatory factor 4 |
Gene Aliases: |
LSIRF, MUM1, NF-EM5, SHEP8 |
Location: |
Chr.6:391739-411443 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| IRF4 | NM_001195286.1 | 9 | 2065 | NP_001182215.1 |
| NM_002460.3 | 9 | 2068 | NP_002451.2 | |
| NR_046000.2 | 10 | 2199 | ||
| XM_006715090.2 | 8 | 1947 | XP_006715153.1 | |
| BC015752.1 | 9 | 2015 | AAH15752.1 | |
| U52682.1 | 9 | 2064 | AAC50779.1 | |
| U63738.1 | 9 | 2047 | AAB37258.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv830569 | Chr.6:402743 - 573991 on Build GRCh38 | Gain |
|
| nsv600780 | Chr.6:403799 - 442410 on Build GRCh38 | Loss |
|
| nsv821644 | Chr.6:182761 - 411478 on Build GRCh38 | Gain |
|
| nsv830568 | Chr.6:232604 - 411249 on Build GRCh38 | Gain+Loss |
|
| esv3607852 | Chr.6:381951 - 500339 on Build GRCh38 | Loss |
|
| nsv600779 | Chr.6:383546 - 439985 on Build GRCh38 | Loss |
|
| esv3607835 | Chr.6:180946 - 734275 on Build GRCh38 | Loss |
|
| nsv823390 | Chr.6:295899 - 452604 on Build GRCh38 | Gain |
|
| dgv5884n100 | Chr.6:257341 - 447036 on Build GRCh38 | Gain |
|
| nsv509115 | Chr.6:360763 - 416001 on Build GRCh38 | Insertion |
|
| nsv428134 | Chr.6:60001 - 653718 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map