Assay Details
Target Gene Details
Entrez Gene ID: | 5521 |
Gene Name: | protein phosphatase 2 regulatory subunit Bbeta |
Gene Aliases: |
B55BETA, PP2AB55BETA, PP2ABBETA, PP2APR55B, PP2APR55BETA, PR2AB55BETA, PR2ABBETA, PR2APR55BETA, PR52B, PR55-BETA, PR55BETA, SCA12 |
Location: |
Chr.5:146589504-147081520 on Build GRCh38 |
Assay Gene Location: | Within Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PPP2R2B | NM_001271899.1 | 10 | 1551 | NP_001258828.1 |
| NM_001271900.1 | 11 | 1745 | NP_001258829.1 | |
| NM_001271948.1 | 10 | 2078 | NP_001258877.1 | |
| NM_181674.2 | 10 | 1876 | NP_858060.2 | |
| NM_181675.3 | 10 | 1598 | NP_858061.2 | |
| NM_181676.2 | 9 | 1682 | NP_858062.1 | |
| NM_181677.2 | 11 | 1508 | NP_858063.1 | |
| NM_181678.2 | 11 | 1521 | NP_858064.1 | |
| NR_073526.1 | 9 | 1993 | ||
| NR_073527.1 | 10 | 1630 | ||
| AI621232.1 | 1 | 501 | ||
| AK056192.1 | 10 | 1558 | ||
| AK289717.1 | 10 | 1527 | ||
| AK294659.1 | 11 | 1654 | ||
| AK295347.1 | 10 | 1522 | ||
| AK314810.1 | ||||
| BC031790.1 | 10 | 1766 | AAH31790.1 | |
| BX647887.1 | 10 | 1612 | ||
| M64930.1 | 9 | 1804 | AAA36491.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv5052 | Chr.5:146559163 - 146592780 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map