Assay Details
Target Gene Details
Entrez Gene ID: | 7841 |
Gene Name: | mannosyl-oligosaccharide glucosidase |
Gene Aliases: |
CDG2B, CWH41, DER7, GCS1 |
Location: |
Chr.2:74461057-74465410 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MOGS | NM_001146158.1 | 5 | 1169 | NP_001139630.1 |
| NM_006302.2 | 4 | 1495 | NP_006293.2 | |
| XM_017004876.1 | 3 | 1258 | XP_016860365.1 | |
| XM_017004877.1 | 4 | 933 | XP_016860366.1 | |
| AK292553.1 | 4 | 1441 | ||
| AK292719.1 | 2 | 1013 | ||
| AK307991.1 | 5 | 1155 | ||
| AK313355.1 | 4 | 1495 | ||
| BC028337.1 | 4 | 1476 | AAH28337.1 | |
| BC117250.1 | 4 | 1368 | ||
| BC117252.1 | 4 | 1368 | ||
| X87237.1 | 4 | 1464 | CAA60683.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv458185 | Chr.2:74367887 - 74531807 on Build GRCh38 | Loss |
|
| esv2759061 | Chr.2:74252797 - 74535298 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map