Assay Details
Target Gene Details
Entrez Gene ID: | 727 |
Gene Name: | complement component 5 |
Gene Aliases: |
C5D, C5a, C5b, CPAMD4, ECLZB |
Location: |
Chr.9:120952335-121075174 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 33 - Exon 34 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| C5 | NM_001317163.1 | NP_001304092.1 | ||
| NM_001735.2 | NP_001726.2 | |||
| XM_011518980.2 | XP_011517282.1 | |||
| XM_017015102.1 | XP_016870591.1 | |||
| XM_017015103.1 | XP_016870592.1 | |||
| AB209031.1 | BAD92268.1 | |||
| BC113738.1 | ||||
| BC113740.1 | ||||
| M57729.1 | AAA51925.1 | |||
| M65134.1 | AAA51856.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2759713 | Chr.9:120885919 - 121080717 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map