Assay Details
Target Gene Details
Entrez Gene ID: | 2070 |
Gene Name: | EYA transcriptional coactivator and phosphatase 4 |
Gene Aliases: |
CMD1J, DFNA10 |
Location: |
Chr.6:133240340-133532120 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 20 - Intron 20 |
Target Gene Details
Entrez Gene ID: | 100507308 |
Gene Name: | TCF21 antisense RNA inducing promoter demethylation |
Gene Aliases: |
EYA4-AS1 |
Location: |
Chr.6:133502252-133889006 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TARID | NR_109982.1 |
More Information
Set Membership: |
|

Genomic Map