Assay Details
Target Gene Details
Entrez Gene ID: | 4982 |
Gene Name: | TNF receptor superfamily member 11b |
Gene Aliases: |
OCIF, OPG, PDB5, TR1 |
Location: |
Chr.8:118923557-118952144 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TNFRSF11B | NM_002546.3 | 5 | 1502 | NP_002537.3 |
| AB002146.1 | BAA25910.1 | |||
| AF134187.1 | 4 | 1117 | AAF20168.1 | |
| AK223155.1 | 5 | 1241 | BAD96875.1 | |
| AK313710.1 | ||||
| BC030155.2 | 5 | 1400 | AAH30155.1 | |
| U94332.1 | 5 | 1273 | AAB53709.1 |
More Information
Set Membership: |
|

Genomic Map