Assay Details
Target Gene Details
Entrez Gene ID: | 9990 |
Gene Name: | solute carrier family 12 member 6 |
Gene Aliases: |
ACCPN, KCC3, KCC3A, KCC3B |
Location: |
Chr.15:34229996-34338064 on Build GRCh38 |
Assay Gene Location: | Within Exon 27 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC12A6 | NM_001042494.1 | 26 | 4947 | NP_001035959.1 |
| NM_001042495.1 | 26 | 4622 | NP_001035960.1 | |
| NM_001042496.1 | 26 | 5013 | NP_001035961.1 | |
| NM_001042497.1 | 24 | 4716 | NP_001035962.1 | |
| NM_005135.2 | 25 | 4586 | NP_005126.1 | |
| NM_133647.1 | 25 | 4761 | NP_598408.1 | |
| XM_006720793.3 | 24 | 5496 | XP_006720856.1 | |
| AK056019.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1052620 | Chr.15:32612402 - 34538731 on Build GRCh38 | Gain |
|
| esv33337 | Chr.15:31689863 - 34575097 on Build GRCh38 | Gain+Loss |
|
| esv3569431 | Chr.15:33564570 - 35080307 on Build GRCh38 | Gain |
|
| dgv4456n54 | Chr.15:32604149 - 34557061 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs117846663] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map