Assay Details
Target Gene Details
Entrez Gene ID: | 57167 |
Gene Name: | spalt like transcription factor 4 |
Gene Aliases: |
DRRS, HSAL4, ZNF797 |
Location: |
Chr.20:51782717-51802523 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SALL4 | NM_001318031.1 | 2 | 1096 | NP_001304960.1 |
| NM_020436.4 | 2 | 1096 | NP_065169.1 | |
| XM_005260467.3 | 2 | 1077 | XP_005260524.1 | |
| XM_011528921.2 | 4 | 1272 | XP_011527223.1 | |
| XM_011528922.2 | 3 | 1231 | XP_011527224.1 | |
| AK001666.1 | 2 | 1048 | ||
| AY170621.1 | 2 | 981 | AAO16566.1 | |
| AY170622.1 | AAO16567.1 | |||
| AY172738.1 | 2 | 981 | AAO44950.1 | |
| BC111714.1 | 2 | 1037 | AAI11715.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv979442 | Chr.20:51790944 - 51792166 on Build GRCh38 | Duplication |
|
| esv2758800 | Chr.20:51706078 - 51802856 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map