Assay Details
Target Gene Details
Entrez Gene ID: | 5027 |
Gene Name: | purinergic receptor P2X 7 |
Gene Aliases: |
P2X7 |
Location: |
Chr.12:121132819-121189478 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| P2RX7 | NM_002562.5 | 1 | 197 | NP_002553.3 |
| NR_033948.1 | 1 | 197 | ||
| NR_033949.1 | 1 | 197 | ||
| NR_033950.1 | 1 | 197 | ||
| NR_033951.1 | 1 | 197 | ||
| NR_033952.1 | 1 | 197 | ||
| NR_033953.1 | 1 | 206 | ||
| NR_033954.1 | 1 | 197 | ||
| NR_033955.1 | 1 | 197 | ||
| NR_033956.1 | 1 | 197 | ||
| XM_011538419.2 | 1 | 206 | XP_011536721.1 | |
| XM_011538420.2 | XP_011536722.1 | |||
| XM_017019364.1 | 1 | 197 | XP_016874853.1 | |
| XM_017019365.1 | 1 | 94 | XP_016874854.1 | |
| XM_017019366.1 | 1 | 138 | XP_016874855.1 | |
| XM_017019367.1 | 1 | 145 | XP_016874856.1 | |
| AB209709.1 | 1 | 118 | BAD92946.1 | |
| AK090866.1 | 1 | 83 | ||
| AK225163.1 | 1 | 83 | ||
| AK290405.1 | 1 | 145 | ||
| AK294126.1 | 1 | 148 | ||
| AK296432.1 | 1 | 145 | ||
| AY847298.1 | 1 | 54 | AAX82087.1 | |
| AY847299.1 | 1 | 54 | AAX82088.1 | |
| AY847300.1 | 1 | 54 | AAX82089.1 | |
| AY847301.1 | 1 | 54 | AAX82090.1 | |
| AY847302.1 | 1 | 54 | AAX82091.1 | |
| AY847303.1 | 1 | 54 | AAX82092.1 | |
| AY847304.1 | 1 | 54 | AAX82093.1 | |
| BC007679.2 | 1 | 95 | AAH07679.1 | |
| BC011913.2 | 1 | 123 | AAH11913.1 | |
| BC121157.1 | 1 | 123 | ||
| BI598073.1 | 1 | 187 | ||
| DA274887.1 | 1 | 83 | ||
| DA302256.1 | 1 | 206 | ||
| DQ399293.1 | 1 | 54 | ||
| GQ180122.1 | 1 | 63 | ||
| Y09561.1 | 1 | 80 | CAA70755.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv560455 | Chr.12:121087788 - 121363295 on Build GRCh38 | Gain |
|
| esv3630934 | Chr.12:121063111 - 121157572 on Build GRCh38 | Loss |
|
| nsv1054878 | Chr.12:121082077 - 121328424 on Build GRCh38 | Gain |
|
| nsv560454 | Chr.12:121019270 - 121307939 on Build GRCh38 | Loss |
|
| nsv521928 | Chr.12:120603925 - 121356945 on Build GRCh38 | Gain |
|
| esv2759917 | Chr.12:120978042 - 121147010 on Build GRCh38 | Loss |
|
| nsv832531 | Chr.12:121002692 - 121166429 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map