Assay Details
Target Gene Details
Entrez Gene ID: | 23054 |
Gene Name: | nuclear receptor coactivator 6 |
Gene Aliases: |
AIB3, ASC2, NRC, PRIP, RAP250, TRBP |
Location: |
Chr.20:34714774-34825649 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 19 - Exon 19 |
More Information
Set Membership: |
|

Genomic Map